Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 120403161-120403434 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSTL1
Entrez Gene ID
11167
Gene Name
follistatin like 1
Gene Aliases
FRP, FSL1, MIR198, OCC-1, OCC1, tsc36
UniGene
Hs.269512
Gene Chromosome Location
Chr.3: 120394214-120451071 on Build GRCh38
Transcripts
NM_007085.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2151759 c.694+1G>A p.? Chr.3: 120403241-120403241 on Build GRCh38
6395032 c.625G>T p.A209S Chr.3: 120403311-120403311 on Build GRCh38
4841581 c.688G>C p.E230Q Chr.3: 120403248-120403248 on Build GRCh38
5990770 c.623_624AT>TG p.N208M Chr.3: 120403312-120403313 on Build GRCh38
728015 c.585A>T p.G195G Chr.3: 120403351-120403351 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1057443156 Chr.3: 120403203-120403203 on Build GRCh38
rs756179625 Chr.3: 120403192-120403192 on Build GRCh38
rs894351664 Chr.3: 120403243-120403243 on Build GRCh38
rs202178483 Chr.3: 120403260-120403260 on Build GRCh38
rs763907992 Chr.3: 120403340-120403340 on Build GRCh38
rs749778932 Chr.3: 120403210-120403210 on Build GRCh38
rs762687164 Chr.3: 120403337-120403337 on Build GRCh38
rs746687531 Chr.3: 120403261-120403261 on Build GRCh38
rs566161223 Chr.3: 120403346-120403346 on Build GRCh38
rs568907330 Chr.3: 120403406-120403406 on Build GRCh38
rs200530287 Chr.3: 120403284-120403284 on Build GRCh38
rs775424514 Chr.3: 120403336-120403336 on Build GRCh38
rs535822904 Chr.3: 120403196-120403196 on Build GRCh38
rs373170332 Chr.3: 120403335-120403335 on Build GRCh38
rs770440542 Chr.3: 120403281-120403281 on Build GRCh38
rs891243439 Chr.3: 120403220-120403220 on Build GRCh38
rs990477959 Chr.3: 120403396-120403396 on Build GRCh38
rs760281818 Chr.3: 120403369-120403369 on Build GRCh38
rs373993322 Chr.3: 120403384-120403384 on Build GRCh38
rs755090190 Chr.3: 120403380-120403380 on Build GRCh38
rs755494176 Chr.3: 120403211-120403211 on Build GRCh38
rs551872681 Chr.3: 120403400-120403400 on Build GRCh38
rs375737073 Chr.3: 120403361-120403361 on Build GRCh38
rs766467366 Chr.3: 120403362-120403362 on Build GRCh38
rs748682716 Chr.3: 120403399-120403399 on Build GRCh38
rs748455860 Chr.3: 120403257-120403257 on Build GRCh38
rs759516293 Chr.3: 120403293-120403293 on Build GRCh38
rs116007134 Chr.3: 120403373-120403373 on Build GRCh38
rs547683717 Chr.3: 120403259-120403259 on Build GRCh38
rs375055685 Chr.3: 120403215-120403215 on Build GRCh38
rs780040341 Chr.3: 120403209-120403209 on Build GRCh38

View Full Product Details