Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 35985649-35985894 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCL14
Entrez Gene ID
6358
Gene Name
C-C motif chemokine ligand 14
Gene Aliases
CC-1, CC-3, CKB1, HCC-1, HCC-1(1-74), HCC-1/HCC-3, HCC-3, MCIF, NCC-2, NCC2, SCYA14, SCYL2, SY14
UniGene
Hs.272493
Gene Chromosome Location
Chr.17: 35983656-35986728 on Build GRCh38
Transcripts
NM_032962.4 NM_032963.3 NR_027921.3 NR_027922.3

Gene Symbol
CCL15-CCL14
Entrez Gene ID
348249
Gene Name
CCL15-CCL14 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.17: 35983656-36002038 on Build GRCh38
Transcripts
NM_032962.4 NM_032963.3 NR_027921.3 NR_027922.3

Gene Symbol
LOC101927339
Entrez Gene ID
101927339
Gene Name
uncharacterized LOC101927339
Gene Aliases
-
UniGene
Hs.569800
Gene Chromosome Location
Chr.17: 35984459-35990270 on Build GRCh38
Transcripts
NM_032962.4 NM_032963.3 NR_027921.3 NR_027922.3

SNP Detail

SNP ID Chromosome Location
rs904447350 Chr.17: 35985719-35985719 on Build GRCh38
rs1000844094 Chr.17: 35985722-35985722 on Build GRCh38
rs745567030 Chr.17: 35985839-35985839 on Build GRCh38
rs1042942517 Chr.17: 35985697-35985697 on Build GRCh38
rs909027611 Chr.17: 35985763-35985763 on Build GRCh38
rs1034116059 Chr.17: 35985776-35985776 on Build GRCh38
rs1038894452 Chr.17: 35985785-35985785 on Build GRCh38
rs756791003 Chr.17: 35985710-35985710 on Build GRCh38
rs1010157870 Chr.17: 35985696-35985696 on Build GRCh38
rs780632165 Chr.17: 35985833-35985833 on Build GRCh38
rs763925611 Chr.17: 35985670-35985670 on Build GRCh38
rs751441115 Chr.17: 35985689-35985689 on Build GRCh38
rs114534185 Chr.17: 35985872-35985872 on Build GRCh38

View Full Product Details