Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 48527128-48527357 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EBP
Entrez Gene ID
10682
Gene Name
emopamil binding protein (sterol isomerase)
Gene Aliases
CDPX2, CHO2, CPX, CPXD, MEND
UniGene
Hs.30619
Gene Chromosome Location
Chr.X: 48521776-48528716 on Build GRCh38
Transcripts
NM_006579.2

SNP Detail

SNP ID Chromosome Location
rs886039345 Chr.X: 48527255-48527255 on Build GRCh38
rs781925754 Chr.X: 48527310-48527310 on Build GRCh38
rs782380458 Chr.X: 48527251-48527251 on Build GRCh38
rs782008789 Chr.X: 48527333-48527333 on Build GRCh38
rs782609400 Chr.X: 48527241-48527241 on Build GRCh38
rs782154327 Chr.X: 48527334-48527334 on Build GRCh38
rs142881014 Chr.X: 48527198-48527198 on Build GRCh38
rs370314537 Chr.X: 48527324-48527324 on Build GRCh38
rs367905193 Chr.X: 48527319-48527319 on Build GRCh38
rs104894792 Chr.X: 48527202-48527202 on Build GRCh38
rs782729537 Chr.X: 48527325-48527325 on Build GRCh38
rs953562416 Chr.X: 48527240-48527240 on Build GRCh38
rs782579338 Chr.X: 48527212-48527212 on Build GRCh38
rs782153209 Chr.X: 48527294-48527294 on Build GRCh38
rs909314502 Chr.X: 48527323-48527323 on Build GRCh38
rs375337168 Chr.X: 48527176-48527176 on Build GRCh38
rs28935174 Chr.X: 48527256-48527256 on Build GRCh38
rs148792902 Chr.X: 48527157-48527157 on Build GRCh38
rs185692210 Chr.X: 48527297-48527297 on Build GRCh38
rs782257058 Chr.X: 48527244-48527244 on Build GRCh38
rs587783615 Chr.X: 48527280-48527281 on Build GRCh38
rs782032266 Chr.X: 48527261-48527261 on Build GRCh38
rs976210101 Chr.X: 48527230-48527230 on Build GRCh38
rs797045545 Chr.X: 48527185-48527195 on Build GRCh38
rs782331959 Chr.X: 48527305-48527305 on Build GRCh38
rs797045546 Chr.X: 48527239-48527243 on Build GRCh38

View Full Product Details