Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 138466693-138466950 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGR1
Entrez Gene ID
1958
Gene Name
early growth response 1
Gene Aliases
AT225, G0S30, KROX-24, NGFI-A, TIS8, ZIF-268, ZNF225
UniGene
Hs.326035
Gene Chromosome Location
Chr.5: 138465492-138469315 on Build GRCh38
Transcripts
NM_001964.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7086669 c.407G>T p.R136L Chr.5: 138466856-138466856 on Build GRCh38
1061389 c.393C>T p.I131I Chr.5: 138466842-138466842 on Build GRCh38
4984018 c.449G>A p.W150* Chr.5: 138466898-138466898 on Build GRCh38
6652330 c.359G>T p.S120I Chr.5: 138466808-138466808 on Build GRCh38
3609004 c.324C>T p.I108I Chr.5: 138466773-138466773 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779279352 Chr.5: 138466783-138466783 on Build GRCh38
rs797019589 Chr.5: 138466862-138466863 on Build GRCh38
rs200402937 Chr.5: 138466757-138466757 on Build GRCh38
rs745933125 Chr.5: 138466841-138466841 on Build GRCh38
rs199802160 Chr.5: 138466734-138466734 on Build GRCh38
rs200134168 Chr.5: 138466813-138466813 on Build GRCh38
rs778216521 Chr.5: 138466812-138466812 on Build GRCh38
rs752604675 Chr.5: 138466711-138466711 on Build GRCh38
rs367940607 Chr.5: 138466730-138466730 on Build GRCh38
rs199627951 Chr.5: 138466835-138466835 on Build GRCh38
rs750013399 Chr.5: 138466908-138466908 on Build GRCh38
rs28365166 Chr.5: 138466881-138466881 on Build GRCh38
rs772577529 Chr.5: 138466811-138466811 on Build GRCh38
rs755545385 Chr.5: 138466764-138466764 on Build GRCh38
rs28365164 Chr.5: 138466884-138466884 on Build GRCh38
rs11743810 Chr.5: 138466715-138466715 on Build GRCh38
rs749875833 Chr.5: 138466751-138466751 on Build GRCh38
rs551889309 Chr.5: 138466785-138466785 on Build GRCh38
rs201028712 Chr.5: 138466721-138466721 on Build GRCh38
rs550624164 Chr.5: 138466795-138466795 on Build GRCh38
rs539786628 Chr.5: 138466866-138466866 on Build GRCh38
rs762815487 Chr.5: 138466886-138466886 on Build GRCh38
rs150291660 Chr.5: 138466786-138466786 on Build GRCh38
rs137878468 Chr.5: 138466900-138466900 on Build GRCh38
rs780705681 Chr.5: 138466747-138466747 on Build GRCh38
rs747264118 Chr.5: 138466827-138466827 on Build GRCh38
rs776813675 Chr.5: 138466836-138466836 on Build GRCh38
rs769829111 Chr.5: 138466847-138466847 on Build GRCh38
rs767122422 Chr.5: 138466903-138466903 on Build GRCh38
rs761499887 Chr.5: 138466902-138466902 on Build GRCh38
rs763911607 Chr.5: 138466893-138466893 on Build GRCh38
rs201694586 Chr.5: 138466828-138466828 on Build GRCh38

View Full Product Details