Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48473941-48474200 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYTH2
Entrez Gene ID
9266
Gene Name
cytohesin 2
Gene Aliases
ARNO, CTS18, CTS18.1, PSCD2, PSCD2L, SEC7L, Sec7p-L, Sec7p-like
UniGene
Hs.144011
Gene Chromosome Location
Chr.19: 48469208-48482314 on Build GRCh38
Transcripts
NM_004228.6 NM_017457.5 XM_006723472.2 XM_006723473.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5770849 c.486G>C p.R162R Chr.19: 48473956-48473956 on Build GRCh38
5878615 c.486G>C p.R162R Chr.19: 48473956-48473956 on Build GRCh38
5390231 c.538C>T p.Q180* Chr.19: 48474008-48474008 on Build GRCh38
998976 c.497C>T p.A166V Chr.19: 48473967-48473967 on Build GRCh38
998978 c.502G>A p.A168T Chr.19: 48473972-48473972 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs28433711 Chr.19: 48474125-48474125 on Build GRCh38
rs755432500 Chr.19: 48474052-48474052 on Build GRCh38
rs765488390 Chr.19: 48474027-48474027 on Build GRCh38
rs766467656 Chr.19: 48474003-48474003 on Build GRCh38
rs1014398047 Chr.19: 48474137-48474137 on Build GRCh38
rs903418691 Chr.19: 48474155-48474155 on Build GRCh38
rs771051877 Chr.19: 48474009-48474009 on Build GRCh38
rs754455899 Chr.19: 48473971-48473971 on Build GRCh38
rs1033420646 Chr.19: 48474164-48474164 on Build GRCh38
rs1010784436 Chr.19: 48474086-48474086 on Build GRCh38
rs773389277 Chr.19: 48474170-48474170 on Build GRCh38
rs377111289 Chr.19: 48473987-48473987 on Build GRCh38
rs780717608 Chr.19: 48473976-48473976 on Build GRCh38
rs546573877 Chr.19: 48474166-48474166 on Build GRCh38
rs937077634 Chr.19: 48474124-48474124 on Build GRCh38
rs907765893 Chr.19: 48474070-48474070 on Build GRCh38
rs780584269 Chr.19: 48474056-48474056 on Build GRCh38
rs771977589 Chr.19: 48474152-48474152 on Build GRCh38
rs556212908 Chr.19: 48474037-48474037 on Build GRCh38
rs755760100 Chr.19: 48473992-48473992 on Build GRCh38
rs755790296 Chr.19: 48474068-48474068 on Build GRCh38
rs567141309 Chr.19: 48474134-48474134 on Build GRCh38
rs898858476 Chr.19: 48474114-48474114 on Build GRCh38
rs544309668 Chr.19: 48474094-48474094 on Build GRCh38
rs940629100 Chr.19: 48474083-48474083 on Build GRCh38
rs188841092 Chr.19: 48474023-48474023 on Build GRCh38
rs181597580 Chr.19: 48474156-48474156 on Build GRCh38
rs769956837 Chr.19: 48474169-48474169 on Build GRCh38
rs975824182 Chr.19: 48474177-48474177 on Build GRCh38
rs774399359 Chr.19: 48474021-48474021 on Build GRCh38
rs562697189 Chr.19: 48474100-48474100 on Build GRCh38
rs776648598 Chr.19: 48474019-48474020 on Build GRCh38
rs766914095 Chr.19: 48474034-48474034 on Build GRCh38
rs759505371 Chr.19: 48474022-48474022 on Build GRCh38
rs372095603 Chr.19: 48474174-48474174 on Build GRCh38
rs1006139156 Chr.19: 48473967-48473967 on Build GRCh38
rs895557450 Chr.19: 48474130-48474130 on Build GRCh38
rs752185512 Chr.19: 48474065-48474065 on Build GRCh38
rs1020483249 Chr.19: 48474140-48474140 on Build GRCh38
rs148173211 Chr.19: 48473989-48473989 on Build GRCh38
rs369066635 Chr.19: 48474026-48474026 on Build GRCh38
rs763279556 Chr.19: 48474030-48474030 on Build GRCh38
rs537835463 Chr.19: 48473979-48473979 on Build GRCh38
rs548442097 Chr.19: 48474133-48474133 on Build GRCh38
rs749092477 Chr.19: 48474006-48474006 on Build GRCh38
rs532630301 Chr.19: 48474029-48474029 on Build GRCh38
rs745813686 Chr.19: 48474138-48474138 on Build GRCh38

View Full Product Details