Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 7563611-7563872 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSP
Entrez Gene ID
1832
Gene Name
desmoplakin
Gene Aliases
DCWHKTA, DP
UniGene
Hs.519873
Gene Chromosome Location
Chr.6: 7541575-7586717 on Build GRCh38
Transcripts
NM_001008844.2 NM_001319034.1 NM_004415.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3162601 c.759G>T p.E253D Chr.6: 7563768-7563768 on Build GRCh38
1081616 c.740C>T p.A247V Chr.6: 7563749-7563749 on Build GRCh38
5822502 c.762G>A p.E254E Chr.6: 7563771-7563771 on Build GRCh38
3922147 c.757G>A p.E253K Chr.6: 7563766-7563766 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs375327581 Chr.6: 7563726-7563726 on Build GRCh38
rs747967791 Chr.6: 7563836-7563836 on Build GRCh38
rs764388232 Chr.6: 7563798-7563798 on Build GRCh38
rs267601117 Chr.6: 7563739-7563739 on Build GRCh38
rs147178098 Chr.6: 7563808-7563808 on Build GRCh38
rs772921319 Chr.6: 7563756-7563756 on Build GRCh38
rs886061745 Chr.6: 7563725-7563725 on Build GRCh38
rs768577244 Chr.6: 7563830-7563830 on Build GRCh38
rs2806234 Chr.6: 7563750-7563750 on Build GRCh38
rs967650168 Chr.6: 7563683-7563689 on Build GRCh38
rs543053921 Chr.6: 7563651-7563651 on Build GRCh38
rs753228677 Chr.6: 7563783-7563783 on Build GRCh38
rs897237729 Chr.6: 7563799-7563799 on Build GRCh38
rs769393246 Chr.6: 7563837-7563837 on Build GRCh38
rs147522043 Chr.6: 7563781-7563781 on Build GRCh38
rs749504597 Chr.6: 7563702-7563702 on Build GRCh38
rs760502491 Chr.6: 7563737-7563737 on Build GRCh38
rs373456357 Chr.6: 7563704-7563704 on Build GRCh38
rs140348839 Chr.6: 7563815-7563815 on Build GRCh38
rs980731625 Chr.6: 7563824-7563824 on Build GRCh38
rs12205970 Chr.6: 7563668-7563668 on Build GRCh38
rs756608550 Chr.6: 7563791-7563791 on Build GRCh38
rs776292608 Chr.6: 7563833-7563833 on Build GRCh38
rs745995077 Chr.6: 7563812-7563812 on Build GRCh38
rs779104522 Chr.6: 7563809-7563809 on Build GRCh38
rs754174261 Chr.6: 7563805-7563805 on Build GRCh38
rs76189583 Chr.6: 7563843-7563843 on Build GRCh38
rs745883736 Chr.6: 7563713-7563713 on Build GRCh38
rs780067594 Chr.6: 7563814-7563814 on Build GRCh38
rs762541786 Chr.6: 7563774-7563774 on Build GRCh38
rs778873989 Chr.6: 7563711-7563711 on Build GRCh38
rs1008042360 Chr.6: 7563797-7563797 on Build GRCh38
rs544360130 Chr.6: 7563749-7563749 on Build GRCh38
rs775383578 Chr.6: 7563729-7563729 on Build GRCh38
rs561918314 Chr.6: 7563698-7563698 on Build GRCh38
rs1049595577 Chr.6: 7563703-7563703 on Build GRCh38
rs372016373 Chr.6: 7563701-7563701 on Build GRCh38
rs141028505 Chr.6: 7563738-7563738 on Build GRCh38

View Full Product Details