Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 94054953-94055226 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX24
Entrez Gene ID
57062
Gene Name
DEAD-box helicase 24
Gene Aliases
-
UniGene
Hs.510328
Gene Chromosome Location
Chr.14: 94050922-94081212 on Build GRCh38
Transcripts
NM_020414.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1493341 c.2147C>T p.P716L Chr.14: 94055027-94055027 on Build GRCh38
4676788 c.2122A>G p.K708E Chr.14: 94055052-94055052 on Build GRCh38
5578383 c.2146C>T p.P716S Chr.14: 94055028-94055028 on Build GRCh38
1707798 c.2119A>T p.K707* Chr.14: 94055055-94055055 on Build GRCh38
7140490 c.2118C>A p.L706L Chr.14: 94055056-94055056 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770109956 Chr.14: 94055193-94055193 on Build GRCh38
rs772955552 Chr.14: 94055200-94055200 on Build GRCh38
rs762986578 Chr.14: 94055002-94055002 on Build GRCh38
rs771827163 Chr.14: 94055197-94055197 on Build GRCh38
rs370120289 Chr.14: 94055143-94055143 on Build GRCh38
rs760419383 Chr.14: 94055141-94055141 on Build GRCh38
rs778124716 Chr.14: 94055070-94055070 on Build GRCh38
rs755607741 Chr.14: 94055032-94055032 on Build GRCh38
rs747280169 Chr.14: 94055072-94055072 on Build GRCh38
rs748133717 Chr.14: 94055189-94055189 on Build GRCh38
rs747408850 Chr.14: 94055171-94055171 on Build GRCh38
rs763968440 Chr.14: 94055010-94055010 on Build GRCh38
rs149275488 Chr.14: 94055034-94055034 on Build GRCh38
rs754272738 Chr.14: 94055042-94055042 on Build GRCh38
rs761593879 Chr.14: 94055102-94055102 on Build GRCh38
rs765766735 Chr.14: 94055148-94055148 on Build GRCh38
rs781494399 Chr.14: 94055038-94055038 on Build GRCh38
rs771096477 Chr.14: 94054982-94054982 on Build GRCh38
rs548009470 Chr.14: 94055026-94055026 on Build GRCh38
rs553018329 Chr.14: 94055101-94055101 on Build GRCh38
rs201027983 Chr.14: 94055183-94055183 on Build GRCh38
rs897721611 Chr.14: 94055184-94055184 on Build GRCh38
rs753310673 Chr.14: 94055150-94055150 on Build GRCh38
rs755474561 Chr.14: 94055059-94055059 on Build GRCh38
rs761612140 Chr.14: 94055017-94055017 on Build GRCh38
rs776731853 Chr.14: 94055093-94055093 on Build GRCh38
rs758808678 Chr.14: 94055153-94055153 on Build GRCh38
rs747489286 Chr.14: 94055025-94055025 on Build GRCh38
rs895475296 Chr.14: 94055104-94055104 on Build GRCh38
rs768579258 Chr.14: 94055097-94055097 on Build GRCh38
rs532958033 Chr.14: 94055158-94055158 on Build GRCh38
rs779313699 Chr.14: 94055060-94055060 on Build GRCh38
rs925724142 Chr.14: 94055077-94055077 on Build GRCh38
rs185805781 Chr.14: 94055122-94055122 on Build GRCh38
rs775268046 Chr.14: 94054983-94054983 on Build GRCh38
rs994685084 Chr.14: 94055201-94055201 on Build GRCh38
rs748447688 Chr.14: 94055076-94055077 on Build GRCh38
rs748455375 Chr.14: 94055063-94055063 on Build GRCh38
rs778125782 Chr.14: 94055014-94055014 on Build GRCh38
rs772829748 Chr.14: 94055135-94055135 on Build GRCh38
rs145305591 Chr.14: 94055095-94055095 on Build GRCh38
rs770885223 Chr.14: 94055088-94055088 on Build GRCh38
rs191501585 Chr.14: 94055182-94055182 on Build GRCh38
rs745895108 Chr.14: 94055179-94055179 on Build GRCh38
rs772628573 Chr.14: 94055069-94055069 on Build GRCh38
rs757763942 Chr.14: 94055029-94055029 on Build GRCh38
rs754448577 Chr.14: 94055147-94055147 on Build GRCh38
rs753342303 Chr.14: 94055145-94055145 on Build GRCh38
rs369246793 Chr.14: 94055173-94055173 on Build GRCh38
rs746253081 Chr.14: 94055058-94055058 on Build GRCh38
rs749273528 Chr.14: 94055070-94055072 on Build GRCh38
rs781281886 Chr.14: 94055178-94055178 on Build GRCh38
rs767359462 Chr.14: 94055126-94055126 on Build GRCh38

View Full Product Details