Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 77805796-77806037 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLG5
Entrez Gene ID
9231
Gene Name
discs large MAGUK scaffold protein 5
Gene Aliases
LP-DLG, P-DLG5, PDLG
UniGene
Hs.652690
Gene Chromosome Location
Chr.10: 77790787-77926590 on Build GRCh38
Transcripts
NM_004747.3 XM_005270276.4 XM_006718056.3 XM_011540341.2 XM_011540342.1 XM_011540344.2 XM_011540345.1 XM_011540346.2 XM_011540347.2 XM_017016913.1 XM_017016914.1 XM_017016915.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6424268 c.5024G>T p.S1675I Chr.10: 77805805-77805805 on Build GRCh38
5574564 c.5019C>A p.D1673E Chr.10: 77805810-77805810 on Build GRCh38
6461361 c.5016T>C p.D1672D Chr.10: 77805813-77805813 on Build GRCh38
3440869 c.4995C>T p.L1665L Chr.10: 77805834-77805834 on Build GRCh38
3770695 c.5029A>G p.T1677A Chr.10: 77805800-77805800 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761585268 Chr.10: 77805889-77805889 on Build GRCh38
rs768409732 Chr.10: 77805896-77805896 on Build GRCh38
rs772792300 Chr.10: 77805910-77805918 on Build GRCh38
rs932723417 Chr.10: 77805986-77805986 on Build GRCh38
rs746635695 Chr.10: 77805888-77805888 on Build GRCh38
rs575052345 Chr.10: 77805960-77805960 on Build GRCh38
rs149797176 Chr.10: 77805844-77805844 on Build GRCh38
rs944444489 Chr.10: 77805891-77805891 on Build GRCh38
rs185398005 Chr.10: 77806011-77806011 on Build GRCh38
rs761351074 Chr.10: 77805901-77805901 on Build GRCh38
rs779352223 Chr.10: 77805830-77805830 on Build GRCh38
rs764014393 Chr.10: 77806011-77806011 on Build GRCh38
rs996999063 Chr.10: 77805963-77805963 on Build GRCh38
rs944501025 Chr.10: 77805955-77805955 on Build GRCh38
rs1049859026 Chr.10: 77806010-77806010 on Build GRCh38
rs565102177 Chr.10: 77805933-77805933 on Build GRCh38
rs796704852 Chr.10: 77805925-77805925 on Build GRCh38
rs746396118 Chr.10: 77805839-77805839 on Build GRCh38
rs757672815 Chr.10: 77805828-77805828 on Build GRCh38
rs904750200 Chr.10: 77805999-77805999 on Build GRCh38
rs563053873 Chr.10: 77805966-77805966 on Build GRCh38
rs181618464 Chr.10: 77805971-77805971 on Build GRCh38
rs530075256 Chr.10: 77805998-77805998 on Build GRCh38
rs935705165 Chr.10: 77805882-77805882 on Build GRCh38
rs2289311 Chr.10: 77805897-77805897 on Build GRCh38
rs911568082 Chr.10: 77805870-77805870 on Build GRCh38
rs769972568 Chr.10: 77805825-77805825 on Build GRCh38
rs370462253 Chr.10: 77806012-77806012 on Build GRCh38
rs775833263 Chr.10: 77805872-77805872 on Build GRCh38

View Full Product Details