Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 128515418-128515691 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLE1
Entrez Gene ID
2733
Gene Name
GLE1, RNA export mediator
Gene Aliases
GLE1L, LCCS, LCCS1, hGLE1
UniGene
Hs.522418
Gene Chromosome Location
Chr.9: 128504692-128542301 on Build GRCh38
Transcripts
NM_001003722.1 NM_001499.2 XM_006717059.2 XM_006717060.2 XM_011518549.1 XM_011518550.1 XM_011518551.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
455415 c.361C>T p.Q121* Chr.9: 128515568-128515568 on Build GRCh38
6422964 c.340C>A p.H114N Chr.9: 128515547-128515547 on Build GRCh38
6422963 c.340C>A p.H114N Chr.9: 128515547-128515547 on Build GRCh38
4921666 c.371G>T p.R124L Chr.9: 128515578-128515578 on Build GRCh38
4921667 c.371G>T p.R124L Chr.9: 128515578-128515578 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770160699 Chr.9: 128515484-128515484 on Build GRCh38
rs763162057 Chr.9: 128515522-128515522 on Build GRCh38
rs768968019 Chr.9: 128515591-128515591 on Build GRCh38
rs17852725 Chr.9: 128515596-128515596 on Build GRCh38
rs775737974 Chr.9: 128515485-128515485 on Build GRCh38
rs1043432125 Chr.9: 128515494-128515494 on Build GRCh38
rs751329882 Chr.9: 128515563-128515563 on Build GRCh38
rs775067389 Chr.9: 128515607-128515607 on Build GRCh38
rs530867627 Chr.9: 128515601-128515601 on Build GRCh38
rs774315243 Chr.9: 128515656-128515657 on Build GRCh38
rs548254579 Chr.9: 128515530-128515530 on Build GRCh38
rs755241713 Chr.9: 128515654-128515654 on Build GRCh38
rs763389420 Chr.9: 128515490-128515491 on Build GRCh38
rs898521572 Chr.9: 128515457-128515457 on Build GRCh38
rs753795376 Chr.9: 128515642-128515642 on Build GRCh38
rs763940805 Chr.9: 128515561-128515561 on Build GRCh38
rs143432034 Chr.9: 128515639-128515639 on Build GRCh38
rs769043154 Chr.9: 128515493-128515493 on Build GRCh38
rs1026190802 Chr.9: 128515476-128515476 on Build GRCh38
rs779059573 Chr.9: 128515664-128515664 on Build GRCh38
rs200493861 Chr.9: 128515497-128515497 on Build GRCh38
rs774777393 Chr.9: 128515499-128515499 on Build GRCh38
rs370361962 Chr.9: 128515600-128515600 on Build GRCh38
rs756230035 Chr.9: 128515578-128515578 on Build GRCh38
rs772397206 Chr.9: 128515667-128515667 on Build GRCh38
rs913143540 Chr.9: 128515471-128515471 on Build GRCh38
rs373915964 Chr.9: 128515613-128515613 on Build GRCh38
rs137939042 Chr.9: 128515534-128515534 on Build GRCh38
rs769327507 Chr.9: 128515605-128515605 on Build GRCh38
rs761658624 Chr.9: 128515617-128515617 on Build GRCh38
rs528472377 Chr.9: 128515525-128515525 on Build GRCh38
rs762303888 Chr.9: 128515513-128515513 on Build GRCh38
rs568237790 Chr.9: 128515560-128515560 on Build GRCh38
rs374485439 Chr.9: 128515670-128515670 on Build GRCh38
rs757269933 Chr.9: 128515564-128515564 on Build GRCh38
rs147114045 Chr.9: 128515528-128515528 on Build GRCh38
rs369524210 Chr.9: 128515587-128515587 on Build GRCh38
rs1056901199 Chr.9: 128515446-128515446 on Build GRCh38
rs986607301 Chr.9: 128515622-128515622 on Build GRCh38
rs750559006 Chr.9: 128515568-128515568 on Build GRCh38
rs377567854 Chr.9: 128515612-128515612 on Build GRCh38
rs372645952 Chr.9: 128515494-128515500 on Build GRCh38
rs749653692 Chr.9: 128515488-128515488 on Build GRCh38
rs748454529 Chr.9: 128515598-128515598 on Build GRCh38
rs762710330 Chr.9: 128515549-128515549 on Build GRCh38
rs890549653 Chr.9: 128515569-128515569 on Build GRCh38
rs745881176 Chr.9: 128515480-128515480 on Build GRCh38
rs938495139 Chr.9: 128515442-128515442 on Build GRCh38
rs780186647 Chr.9: 128515583-128515583 on Build GRCh38
rs374673335 Chr.9: 128515604-128515604 on Build GRCh38
rs750362620 Chr.9: 128515635-128515635 on Build GRCh38
rs904550265 Chr.9: 128515517-128515517 on Build GRCh38
rs202032635 Chr.9: 128515496-128515496 on Build GRCh38
rs752367103 Chr.9: 128515545-128515545 on Build GRCh38
rs781224143 Chr.9: 128515565-128515565 on Build GRCh38
rs748544571 Chr.9: 128515665-128515665 on Build GRCh38
rs767599399 Chr.9: 128515618-128515618 on Build GRCh38

View Full Product Details