Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 41226069-41226342 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BACE2
Entrez Gene ID
25825
Gene Name
beta-site APP-cleaving enzyme 2
Gene Aliases
AEPLC, ALP56, ASP1, ASP21, BAE2, CDA13, CEAP1, DRAP
UniGene
Hs.43047
Gene Chromosome Location
Chr.21: 41167801-41282534 on Build GRCh38
Transcripts
NM_012105.4 NM_138991.2 NM_138992.2 XM_017028314.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4843702 c.328G>T p.D110Y Chr.21: 41226281-41226281 on Build GRCh38
6452879 c.352G>A p.V118M Chr.21: 41226305-41226305 on Build GRCh38
6216191 c.355G>A p.A119T Chr.21: 41226308-41226308 on Build GRCh38
396748 c.386A>G p.Y129C Chr.21: 41226339-41226339 on Build GRCh38
4664980 c.342T>C p.S114S Chr.21: 41226295-41226295 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772712218 Chr.21: 41226231-41226231 on Build GRCh38
rs763198165 Chr.21: 41226238-41226238 on Build GRCh38
rs368254374 Chr.21: 41226223-41226223 on Build GRCh38
rs564358286 Chr.21: 41226142-41226143 on Build GRCh38
rs57826923 Chr.21: 41226142-41226142 on Build GRCh38
rs904304165 Chr.21: 41226214-41226214 on Build GRCh38
rs753326379 Chr.21: 41226237-41226238 on Build GRCh38
rs376279963 Chr.21: 41226229-41226229 on Build GRCh38
rs370446164 Chr.21: 41226237-41226237 on Build GRCh38
rs754811935 Chr.21: 41226285-41226285 on Build GRCh38
rs796190034 Chr.21: 41226150-41226150 on Build GRCh38
rs960938707 Chr.21: 41226218-41226218 on Build GRCh38
rs765438542 Chr.21: 41226251-41226251 on Build GRCh38
rs776977651 Chr.21: 41226247-41226247 on Build GRCh38
rs761168357 Chr.21: 41226243-41226243 on Build GRCh38
rs58677403 Chr.21: 41226150-41226151 on Build GRCh38
rs967991610 Chr.21: 41226190-41226190 on Build GRCh38
rs149411639 Chr.21: 41226258-41226258 on Build GRCh38
rs751479561 Chr.21: 41226278-41226278 on Build GRCh38
rs372472410 Chr.21: 41226305-41226305 on Build GRCh38
rs796140954 Chr.21: 41226141-41226142 on Build GRCh38
rs978045989 Chr.21: 41226196-41226196 on Build GRCh38
rs923974669 Chr.21: 41226197-41226197 on Build GRCh38
rs745860170 Chr.21: 41226154-41226154 on Build GRCh38
rs777419414 Chr.21: 41226318-41226318 on Build GRCh38
rs368473609 Chr.21: 41226304-41226304 on Build GRCh38
rs17000685 Chr.21: 41226179-41226179 on Build GRCh38
rs772583182 Chr.21: 41226226-41226226 on Build GRCh38
rs764522009 Chr.21: 41226245-41226245 on Build GRCh38
rs750471427 Chr.21: 41226259-41226259 on Build GRCh38
rs553756921 Chr.21: 41226132-41226132 on Build GRCh38
rs766328992 Chr.21: 41226277-41226277 on Build GRCh38
rs778974920 Chr.21: 41226116-41226116 on Build GRCh38
rs758429022 Chr.21: 41226268-41226268 on Build GRCh38
rs755735530 Chr.21: 41226315-41226315 on Build GRCh38
rs374082955 Chr.21: 41226248-41226248 on Build GRCh38
rs748840037 Chr.21: 41226319-41226319 on Build GRCh38
rs534008804 Chr.21: 41226131-41226131 on Build GRCh38
rs761924059 Chr.21: 41226249-41226249 on Build GRCh38

View Full Product Details