Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7508037-7508298 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf45
Entrez Gene ID
374877
Gene Name
chromosome 19 open reading frame 45
Gene Aliases
-
UniGene
Hs.631862
Gene Chromosome Location
Chr.19: 7497559-7508457 on Build GRCh38
Transcripts
NM_198534.2 XM_005272473.3 XM_011527993.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7426222 c.1313A>T p.K438M Chr.19: 7508267-7508267 on Build GRCh38
7429040 c.1277C>T p.P426L Chr.19: 7508231-7508231 on Build GRCh38
475468 c.1291C>T p.R431C Chr.19: 7508245-7508245 on Build GRCh38
1283756 c.1296C>T p.F432F Chr.19: 7508250-7508250 on Build GRCh38
5810633 c.1337A>T p.Q446L Chr.19: 7508291-7508291 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs915287869 Chr.19: 7508120-7508120 on Build GRCh38
rs897640600 Chr.19: 7508171-7508171 on Build GRCh38
rs149206817 Chr.19: 7508242-7508242 on Build GRCh38
rs368642225 Chr.19: 7508187-7508187 on Build GRCh38
rs761530909 Chr.19: 7508233-7508233 on Build GRCh38
rs758354046 Chr.19: 7508163-7508163 on Build GRCh38
rs878993541 Chr.19: 7508232-7508232 on Build GRCh38
rs778106137 Chr.19: 7508185-7508185 on Build GRCh38
rs371617635 Chr.19: 7508189-7508189 on Build GRCh38
rs753928641 Chr.19: 7508249-7508249 on Build GRCh38
rs988049782 Chr.19: 7508115-7508115 on Build GRCh38
rs776088007 Chr.19: 7508230-7508230 on Build GRCh38
rs746274100 Chr.19: 7508201-7508201 on Build GRCh38
rs916447811 Chr.19: 7508247-7508249 on Build GRCh38
rs765878438 Chr.19: 7508262-7508263 on Build GRCh38
rs1031899312 Chr.19: 7508076-7508076 on Build GRCh38
rs752081628 Chr.19: 7508196-7508196 on Build GRCh38
rs989961188 Chr.19: 7508102-7508102 on Build GRCh38
rs956839522 Chr.19: 7508098-7508098 on Build GRCh38
rs780198790 Chr.19: 7508167-7508167 on Build GRCh38
rs867152684 Chr.19: 7508211-7508211 on Build GRCh38
rs753283645 Chr.19: 7508275-7508279 on Build GRCh38
rs139224067 Chr.19: 7508272-7508272 on Build GRCh38
rs764866109 Chr.19: 7508238-7508238 on Build GRCh38
rs927858529 Chr.19: 7508148-7508148 on Build GRCh38
rs145194803 Chr.19: 7508221-7508221 on Build GRCh38
rs1031567581 Chr.19: 7508081-7508081 on Build GRCh38
rs939207609 Chr.19: 7508152-7508152 on Build GRCh38
rs762267164 Chr.19: 7508259-7508263 on Build GRCh38
rs765091900 Chr.19: 7508157-7508157 on Build GRCh38
rs948020256 Chr.19: 7508121-7508121 on Build GRCh38
rs751582045 Chr.19: 7508246-7508246 on Build GRCh38
rs475923 Chr.19: 7508212-7508212 on Build GRCh38
rs757485099 Chr.19: 7508255-7508255 on Build GRCh38
rs776013435 Chr.19: 7508220-7508220 on Build GRCh38
rs201554277 Chr.19: 7508271-7508271 on Build GRCh38
rs888792115 Chr.19: 7508245-7508245 on Build GRCh38
rs1017803653 Chr.19: 7508115-7508115 on Build GRCh38
rs766389148 Chr.19: 7508244-7508244 on Build GRCh38
rs762891241 Chr.19: 7508243-7508243 on Build GRCh38
rs752803617 Chr.19: 7508177-7508177 on Build GRCh38
rs954868720 Chr.19: 7508109-7508109 on Build GRCh38
rs779329105 Chr.19: 7508198-7508198 on Build GRCh38
rs143413190 Chr.19: 7508268-7508268 on Build GRCh38
rs980665493 Chr.19: 7508131-7508131 on Build GRCh38
rs768177555 Chr.19: 7508227-7508227 on Build GRCh38
rs756305681 Chr.19: 7508179-7508179 on Build GRCh38
rs750261729 Chr.19: 7508158-7508158 on Build GRCh38

View Full Product Details