Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 184577055-184577298 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHB3
Entrez Gene ID
2049
Gene Name
EPH receptor B3
Gene Aliases
EK2, ETK2, HEK2, TYRO6
UniGene
Hs.2913
Gene Chromosome Location
Chr.3: 184561799-184582408 on Build GRCh38
Transcripts
NM_004443.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1617279 c.1326G>A p.A442A Chr.3: 184577155-184577155 on Build GRCh38
730189 c.1227G>T p.R409R Chr.3: 184577056-184577056 on Build GRCh38
4856832 c.1249G>A p.A417T Chr.3: 184577078-184577078 on Build GRCh38
4878509 c.1319G>A p.R440H Chr.3: 184577148-184577148 on Build GRCh38
6164092 c.1255A>G p.T419A Chr.3: 184577084-184577084 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142854760 Chr.3: 184577087-184577087 on Build GRCh38
rs779216599 Chr.3: 184577136-184577136 on Build GRCh38
rs569320067 Chr.3: 184577100-184577100 on Build GRCh38
rs760437107 Chr.3: 184577155-184577155 on Build GRCh38
rs572740428 Chr.3: 184577226-184577226 on Build GRCh38
rs776201525 Chr.3: 184577154-184577154 on Build GRCh38
rs756275485 Chr.3: 184577124-184577124 on Build GRCh38
rs192397531 Chr.3: 184577231-184577231 on Build GRCh38
rs372920731 Chr.3: 184577184-184577184 on Build GRCh38
rs370732996 Chr.3: 184577148-184577148 on Build GRCh38
rs34076689 Chr.3: 184577143-184577143 on Build GRCh38
rs753545856 Chr.3: 184577159-184577159 on Build GRCh38
rs140911993 Chr.3: 184577110-184577110 on Build GRCh38
rs753769374 Chr.3: 184577221-184577221 on Build GRCh38
rs376828208 Chr.3: 184577116-184577116 on Build GRCh38
rs749567734 Chr.3: 184577141-184577141 on Build GRCh38
rs757570445 Chr.3: 184577225-184577225 on Build GRCh38
rs751566289 Chr.3: 184577217-184577217 on Build GRCh38
rs777693513 Chr.3: 184577125-184577125 on Build GRCh38
rs947277288 Chr.3: 184577081-184577081 on Build GRCh38
rs539235549 Chr.3: 184577211-184577211 on Build GRCh38
rs1035366518 Chr.3: 184577247-184577247 on Build GRCh38
rs887962387 Chr.3: 184577201-184577201 on Build GRCh38
rs749280942 Chr.3: 184577224-184577224 on Build GRCh38
rs771706469 Chr.3: 184577092-184577092 on Build GRCh38
rs958677297 Chr.3: 184577264-184577264 on Build GRCh38
rs7644069 Chr.3: 184577220-184577220 on Build GRCh38
rs1044148218 Chr.3: 184577196-184577196 on Build GRCh38
rs369900872 Chr.3: 184577086-184577086 on Build GRCh38
rs375569717 Chr.3: 184577210-184577210 on Build GRCh38
rs951062210 Chr.3: 184577084-184577084 on Build GRCh38
rs1012682713 Chr.3: 184577272-184577272 on Build GRCh38
rs56029711 Chr.3: 184577147-184577147 on Build GRCh38
rs373788594 Chr.3: 184577088-184577088 on Build GRCh38
rs763505156 Chr.3: 184577085-184577085 on Build GRCh38
rs763706707 Chr.3: 184577158-184577158 on Build GRCh38
rs772618334 Chr.3: 184577232-184577232 on Build GRCh38
rs974804741 Chr.3: 184577095-184577095 on Build GRCh38

View Full Product Details