Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 143319682-143319943 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCN1
Entrez Gene ID
1180
Gene Name
chloride voltage-gated channel 1
Gene Aliases
CLC1
UniGene
Hs.121483
Gene Chromosome Location
Chr.7: 143316126-143352008 on Build GRCh38
Transcripts
NM_000083.2 NR_046453.1 XM_017011739.1 XM_017011740.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6370729 c.277G>A p.E93K Chr.7: 143319851-143319851 on Build GRCh38
3762451 c.261C>T p.T87T Chr.7: 143319835-143319835 on Build GRCh38
7083778 c.196A>C p.K66Q Chr.7: 143319770-143319770 on Build GRCh38
6888657 c.252C>T p.S84S Chr.7: 143319826-143319826 on Build GRCh38
4672614 c.181-2A>G p.? Chr.7: 143319753-143319753 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750646253 Chr.7: 143319908-143319908 on Build GRCh38
rs149407148 Chr.7: 143319815-143319815 on Build GRCh38
rs977267957 Chr.7: 143319808-143319808 on Build GRCh38
rs6948526 Chr.7: 143319902-143319902 on Build GRCh38
rs751891017 Chr.7: 143319744-143319744 on Build GRCh38
rs147581794 Chr.7: 143319836-143319836 on Build GRCh38
rs199840356 Chr.7: 143319712-143319712 on Build GRCh38
rs759188441 Chr.7: 143319838-143319838 on Build GRCh38
rs769312894 Chr.7: 143319783-143319783 on Build GRCh38
rs758855079 Chr.7: 143319724-143319724 on Build GRCh38
rs145150180 Chr.7: 143319706-143319706 on Build GRCh38
rs773205379 Chr.7: 143319822-143319822 on Build GRCh38
rs764676032 Chr.7: 143319859-143319859 on Build GRCh38
rs956078618 Chr.7: 143319776-143319776 on Build GRCh38
rs201291713 Chr.7: 143319711-143319711 on Build GRCh38
rs201591839 Chr.7: 143319863-143319863 on Build GRCh38
rs773073006 Chr.7: 143319704-143319705 on Build GRCh38
rs201113237 Chr.7: 143319737-143319737 on Build GRCh38
rs369579634 Chr.7: 143319769-143319769 on Build GRCh38
rs780145291 Chr.7: 143319791-143319791 on Build GRCh38
rs1047548125 Chr.7: 143319916-143319916 on Build GRCh38
rs781383745 Chr.7: 143319761-143319761 on Build GRCh38
rs764582617 Chr.7: 143319887-143319887 on Build GRCh38
rs6962852 Chr.7: 143319835-143319835 on Build GRCh38
rs757543220 Chr.7: 143319750-143319750 on Build GRCh38
rs756334562 Chr.7: 143319909-143319909 on Build GRCh38
rs751982793 Chr.7: 143319898-143319898 on Build GRCh38
rs780034200 Chr.7: 143319917-143319917 on Build GRCh38
rs948634525 Chr.7: 143319840-143319840 on Build GRCh38
rs752230240 Chr.7: 143319861-143319861 on Build GRCh38
rs778162089 Chr.7: 143319735-143319735 on Build GRCh38
rs886062033 Chr.7: 143319789-143319789 on Build GRCh38
rs965601539 Chr.7: 143319701-143319701 on Build GRCh38
rs764736045 Chr.7: 143319717-143319717 on Build GRCh38
rs753174238 Chr.7: 143319721-143319721 on Build GRCh38
rs965481055 Chr.7: 143319781-143319781 on Build GRCh38
rs781557227 Chr.7: 143319906-143319906 on Build GRCh38
rs772100356 Chr.7: 143319818-143319818 on Build GRCh38

View Full Product Details