Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100562770-100563041 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGFG2
Entrez Gene ID
3268
Gene Name
ArfGAP with FG repeats 2
Gene Aliases
HRBL, RABR
UniGene
Hs.521083
Gene Chromosome Location
Chr.7: 100539211-100568220 on Build GRCh38
Transcripts
NM_006076.4 XM_005250306.2 XM_017012112.1 XM_017012114.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1082866 c.1115C>T p.A372V Chr.7: 100562890-100562890 on Build GRCh38
3876333 c.1157A>C p.N386T Chr.7: 100562932-100562932 on Build GRCh38
1622063 c.1152G>A p.Q384Q Chr.7: 100562927-100562927 on Build GRCh38
1622062 c.1116C>T p.A372A Chr.7: 100562891-100562891 on Build GRCh38
3630785 c.1119C>T p.A373A Chr.7: 100562894-100562894 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs916671570 Chr.7: 100562809-100562810 on Build GRCh38
rs376263859 Chr.7: 100562836-100562836 on Build GRCh38
rs746997464 Chr.7: 100562843-100562843 on Build GRCh38
rs754998615 Chr.7: 100562938-100562938 on Build GRCh38
rs146824330 Chr.7: 100562888-100562888 on Build GRCh38
rs764498486 Chr.7: 100562905-100562905 on Build GRCh38
rs538174486 Chr.7: 100562892-100562892 on Build GRCh38
rs750975188 Chr.7: 100562927-100562927 on Build GRCh38
rs370006164 Chr.7: 100562996-100562996 on Build GRCh38
rs373487166 Chr.7: 100562903-100562903 on Build GRCh38
rs772227826 Chr.7: 100562977-100562977 on Build GRCh38
rs780962444 Chr.7: 100562941-100562941 on Build GRCh38
rs745407711 Chr.7: 100562824-100562824 on Build GRCh38
rs747880226 Chr.7: 100562870-100562870 on Build GRCh38
rs186468230 Chr.7: 100562805-100562805 on Build GRCh38
rs541816920 Chr.7: 100562865-100562865 on Build GRCh38
rs762050614 Chr.7: 100562911-100562911 on Build GRCh38
rs375376399 Chr.7: 100562880-100562880 on Build GRCh38
rs765028686 Chr.7: 100562915-100562915 on Build GRCh38
rs757956510 Chr.7: 100562840-100562840 on Build GRCh38
rs777913471 Chr.7: 100562929-100562930 on Build GRCh38
rs965044313 Chr.7: 100562834-100562834 on Build GRCh38
rs775700880 Chr.7: 100562979-100562979 on Build GRCh38
rs372642862 Chr.7: 100562844-100562844 on Build GRCh38
rs373147015 Chr.7: 100562918-100562918 on Build GRCh38
rs758079460 Chr.7: 100562920-100562920 on Build GRCh38
rs996482400 Chr.7: 100562864-100562864 on Build GRCh38
rs200071448 Chr.7: 100562882-100562882 on Build GRCh38
rs777504885 Chr.7: 100562972-100562972 on Build GRCh38
rs756995736 Chr.7: 100562816-100562816 on Build GRCh38
rs34731997 Chr.7: 100562869-100562869 on Build GRCh38
rs746099268 Chr.7: 100562973-100562973 on Build GRCh38
rs897350984 Chr.7: 100562890-100562890 on Build GRCh38
rs1019306102 Chr.7: 100562825-100562825 on Build GRCh38
rs768410184 Chr.7: 100562852-100562852 on Build GRCh38
rs778701263 Chr.7: 100562821-100562821 on Build GRCh38
rs773557903 Chr.7: 100562881-100562881 on Build GRCh38
rs781059535 Chr.7: 100562921-100562921 on Build GRCh38
rs554456777 Chr.7: 100562792-100562792 on Build GRCh38
rs372891507 Chr.7: 100562804-100562804 on Build GRCh38
rs201635709 Chr.7: 100562906-100562906 on Build GRCh38
rs772170595 Chr.7: 100562900-100562900 on Build GRCh38
rs748026655 Chr.7: 100562942-100562942 on Build GRCh38
rs369873014 Chr.7: 100562902-100562902 on Build GRCh38
rs769599892 Chr.7: 100562962-100562962 on Build GRCh38

View Full Product Details