Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 26951248-26951475 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDF1
Entrez Gene ID
126695
Gene Name
keratinocyte differentiation factor 1
Gene Aliases
C1orf172
UniGene
Hs.188881
Gene Chromosome Location
Chr.1: 26949556-26960484 on Build GRCh38
Transcripts
NM_152365.2 XM_005245735.2 XM_011540622.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4667320 c.965G>A p.R322Q Chr.1: 26951416-26951416 on Build GRCh38
7311926 c.961A>T p.T321S Chr.1: 26951420-26951420 on Build GRCh38
5470087 c.982G>A p.A328T Chr.1: 26951399-26951399 on Build GRCh38
6009176 c.1017G>A p.V339V Chr.1: 26951364-26951364 on Build GRCh38
6615133 c.1030A>G p.S344G Chr.1: 26951351-26951351 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774156596 Chr.1: 26951453-26951453 on Build GRCh38
rs746074242 Chr.1: 26951296-26951296 on Build GRCh38
rs780424265 Chr.1: 26951434-26951434 on Build GRCh38
rs780155303 Chr.1: 26951432-26951432 on Build GRCh38
rs774065018 Chr.1: 26951356-26951356 on Build GRCh38
rs747027074 Chr.1: 26951441-26951441 on Build GRCh38
rs758595105 Chr.1: 26951304-26951304 on Build GRCh38
rs374737581 Chr.1: 26951433-26951433 on Build GRCh38
rs750772780 Chr.1: 26951426-26951426 on Build GRCh38
rs150297466 Chr.1: 26951452-26951452 on Build GRCh38
rs370336022 Chr.1: 26951417-26951417 on Build GRCh38
rs767189794 Chr.1: 26951368-26951368 on Build GRCh38
rs202156792 Chr.1: 26951400-26951400 on Build GRCh38
rs769783639 Chr.1: 26951318-26951318 on Build GRCh38
rs1029908828 Chr.1: 26951355-26951355 on Build GRCh38
rs780231971 Chr.1: 26951314-26951314 on Build GRCh38
rs749238119 Chr.1: 26951324-26951324 on Build GRCh38
rs372046904 Chr.1: 26951308-26951308 on Build GRCh38
rs773291830 Chr.1: 26951323-26951323 on Build GRCh38
rs181637936 Chr.1: 26951288-26951288 on Build GRCh38
rs749216836 Chr.1: 26951450-26951450 on Build GRCh38
rs955641715 Chr.1: 26951396-26951396 on Build GRCh38
rs1005612849 Chr.1: 26951291-26951291 on Build GRCh38
rs759180556 Chr.1: 26951362-26951362 on Build GRCh38
rs375717046 Chr.1: 26951373-26951373 on Build GRCh38
rs755094201 Chr.1: 26951446-26951446 on Build GRCh38
rs781487247 Chr.1: 26951449-26951449 on Build GRCh38
rs145055719 Chr.1: 26951425-26951425 on Build GRCh38
rs747116693 Chr.1: 26951316-26951316 on Build GRCh38
rs754368453 Chr.1: 26951408-26951408 on Build GRCh38
rs757746751 Chr.1: 26951415-26951415 on Build GRCh38
rs760195823 Chr.1: 26951399-26951399 on Build GRCh38
rs770752998 Chr.1: 26951339-26951339 on Build GRCh38
rs774148240 Chr.1: 26951388-26951389 on Build GRCh38

View Full Product Details