Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 73386060-73386301 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALMS1
Entrez Gene ID
7840
Gene Name
ALMS1, centrosome and basal body associated protein
Gene Aliases
ALSS
UniGene
Hs.184720
Gene Chromosome Location
Chr.2: 73385758-73609919 on Build GRCh38
Transcripts
NM_015120.4

SNP Detail

SNP ID Chromosome Location
rs760639277 Chr.2: 73386169-73386169 on Build GRCh38
rs956834448 Chr.2: 73386133-73386133 on Build GRCh38
rs899220304 Chr.2: 73386240-73386240 on Build GRCh38
rs886949774 Chr.2: 73386203-73386203 on Build GRCh38
rs771370849 Chr.2: 73386124-73386124 on Build GRCh38
rs764095354 Chr.2: 73386150-73386150 on Build GRCh38
rs767558791 Chr.2: 73386086-73386086 on Build GRCh38
rs1037793096 Chr.2: 73386101-73386101 on Build GRCh38
rs778690416 Chr.2: 73386202-73386202 on Build GRCh38
rs553661392 Chr.2: 73386171-73386171 on Build GRCh38
rs543499940 Chr.2: 73386283-73386283 on Build GRCh38
rs554126571 Chr.2: 73386255-73386255 on Build GRCh38
rs1003701204 Chr.2: 73386214-73386214 on Build GRCh38
rs905373886 Chr.2: 73386087-73386087 on Build GRCh38
rs956562037 Chr.2: 73386228-73386228 on Build GRCh38
rs747750620 Chr.2: 73386113-73386113 on Build GRCh38
rs777177630 Chr.2: 73386140-73386140 on Build GRCh38
rs763023186 Chr.2: 73386149-73386149 on Build GRCh38
rs576081925 Chr.2: 73386256-73386256 on Build GRCh38
rs750326619 Chr.2: 73386151-73386151 on Build GRCh38
rs754611450 Chr.2: 73386183-73386183 on Build GRCh38
rs778271633 Chr.2: 73386120-73386120 on Build GRCh38
rs755847221 Chr.2: 73386096-73386096 on Build GRCh38
rs762635802 Chr.2: 73386086-73386087 on Build GRCh38
rs747490719 Chr.2: 73386223-73386223 on Build GRCh38
rs538598970 Chr.2: 73386146-73386146 on Build GRCh38
rs754662930 Chr.2: 73386103-73386103 on Build GRCh38
rs1031172088 Chr.2: 73386267-73386267 on Build GRCh38
rs1046964700 Chr.2: 73386191-73386191 on Build GRCh38
rs775431837 Chr.2: 73386148-73386148 on Build GRCh38
rs1038210821 Chr.2: 73386234-73386234 on Build GRCh38
rs778264668 Chr.2: 73386109-73386109 on Build GRCh38
rs772450464 Chr.2: 73386117-73386117 on Build GRCh38
rs996611487 Chr.2: 73386248-73386248 on Build GRCh38
rs551575560 Chr.2: 73386222-73386222 on Build GRCh38
rs770203539 Chr.2: 73386147-73386147 on Build GRCh38
rs1025964615 Chr.2: 73386119-73386119 on Build GRCh38
rs11126399 Chr.2: 73386213-73386213 on Build GRCh38
rs989576553 Chr.2: 73386277-73386277 on Build GRCh38
rs890744248 Chr.2: 73386251-73386251 on Build GRCh38
rs187868113 Chr.2: 73386249-73386249 on Build GRCh38
rs747529471 Chr.2: 73386121-73386121 on Build GRCh38
rs868854769 Chr.2: 73386280-73386280 on Build GRCh38
rs1009097793 Chr.2: 73386252-73386252 on Build GRCh38
rs1018027045 Chr.2: 73386160-73386160 on Build GRCh38
rs989191437 Chr.2: 73386134-73386134 on Build GRCh38
rs866402031 Chr.2: 73386095-73386095 on Build GRCh38
rs35414379 Chr.2: 73386274-73386274 on Build GRCh38
rs752293247 Chr.2: 73386209-73386209 on Build GRCh38
rs368947491 Chr.2: 73386102-73386102 on Build GRCh38
rs750332908 Chr.2: 73386089-73386089 on Build GRCh38
rs956885717 Chr.2: 73386272-73386272 on Build GRCh38
rs753680275 Chr.2: 73386178-73386178 on Build GRCh38

View Full Product Details