Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 59619315-59619584 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCNT3
Entrez Gene ID
9245
Gene Name
glucosaminyl (N-acetyl) transferase 3, mucin type
Gene Aliases
C2/4GnT, C24GNT, C2GNT2, C2GNTM, GNTM
UniGene
Hs.194710
Gene Chromosome Location
Chr.15: 59611783-59620011 on Build GRCh38
Transcripts
NM_004751.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4524086 c.1252G>A p.D418N Chr.15: 59619490-59619490 on Build GRCh38
5971480 c.1245G>A p.K415K Chr.15: 59619483-59619483 on Build GRCh38
5892257 c.1276G>A p.E426K Chr.15: 59619514-59619514 on Build GRCh38
4459830 c.1141C>T p.P381S Chr.15: 59619379-59619379 on Build GRCh38
3502826 c.1151G>A p.G384E Chr.15: 59619389-59619389 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11542806 Chr.15: 59619421-59619421 on Build GRCh38
rs201121374 Chr.15: 59619556-59619556 on Build GRCh38
rs773992456 Chr.15: 59619393-59619393 on Build GRCh38
rs756750252 Chr.15: 59619478-59619478 on Build GRCh38
rs1035126847 Chr.15: 59619395-59619396 on Build GRCh38
rs756195924 Chr.15: 59619438-59619438 on Build GRCh38
rs747329705 Chr.15: 59619396-59619396 on Build GRCh38
rs770092791 Chr.15: 59619361-59619361 on Build GRCh38
rs773363536 Chr.15: 59619430-59619430 on Build GRCh38
rs748533221 Chr.15: 59619497-59619497 on Build GRCh38
rs767649062 Chr.15: 59619536-59619536 on Build GRCh38
rs374422549 Chr.15: 59619564-59619564 on Build GRCh38
rs1000368320 Chr.15: 59619367-59619367 on Build GRCh38
rs901250697 Chr.15: 59619557-59619557 on Build GRCh38
rs763611920 Chr.15: 59619504-59619504 on Build GRCh38
rs970423544 Chr.15: 59619432-59619432 on Build GRCh38
rs748810481 Chr.15: 59619377-59619377 on Build GRCh38
rs758640883 Chr.15: 59619348-59619348 on Build GRCh38
rs370534774 Chr.15: 59619545-59619545 on Build GRCh38
rs367791352 Chr.15: 59619463-59619463 on Build GRCh38
rs375933844 Chr.15: 59619524-59619524 on Build GRCh38
rs756078977 Chr.15: 59619370-59619370 on Build GRCh38
rs377505165 Chr.15: 59619471-59619471 on Build GRCh38
rs758008678 Chr.15: 59619562-59619562 on Build GRCh38
rs200137024 Chr.15: 59619499-59619499 on Build GRCh38
rs751791469 Chr.15: 59619460-59619460 on Build GRCh38
rs148598436 Chr.15: 59619558-59619558 on Build GRCh38
rs745569308 Chr.15: 59619394-59619394 on Build GRCh38
rs745884423 Chr.15: 59619358-59619358 on Build GRCh38
rs959110439 Chr.15: 59619408-59619408 on Build GRCh38
rs147773357 Chr.15: 59619400-59619400 on Build GRCh38
rs370136583 Chr.15: 59619523-59619523 on Build GRCh38
rs536974873 Chr.15: 59619466-59619466 on Build GRCh38
rs34081917 Chr.15: 59619412-59619412 on Build GRCh38
rs760459426 Chr.15: 59619546-59619546 on Build GRCh38
rs748094489 Chr.15: 59619365-59619365 on Build GRCh38
rs749920710 Chr.15: 59619561-59619561 on Build GRCh38
rs781488502 Chr.15: 59619446-59619446 on Build GRCh38
rs777185955 Chr.15: 59619336-59619336 on Build GRCh38
rs1025139337 Chr.15: 59619417-59619417 on Build GRCh38
rs778338518 Chr.15: 59619479-59619479 on Build GRCh38
rs750558900 Chr.15: 59619343-59619343 on Build GRCh38
rs771651830 Chr.15: 59619484-59619484 on Build GRCh38
rs769042934 Chr.15: 59619404-59619405 on Build GRCh38
rs780146891 Chr.15: 59619449-59619449 on Build GRCh38
rs199566134 Chr.15: 59619337-59619337 on Build GRCh38
rs770504759 Chr.15: 59619386-59619386 on Build GRCh38
rs745504742 Chr.15: 59619483-59619483 on Build GRCh38
rs765374404 Chr.15: 59619342-59619342 on Build GRCh38
rs781567598 Chr.15: 59619487-59619487 on Build GRCh38
rs773851607 Chr.15: 59619503-59619503 on Build GRCh38
rs147015089 Chr.15: 59619467-59619467 on Build GRCh38
rs777217395 Chr.15: 59619371-59619371 on Build GRCh38
rs372465170 Chr.15: 59619555-59619555 on Build GRCh38
rs763938575 Chr.15: 59619547-59619547 on Build GRCh38
rs776902951 Chr.15: 59619411-59619411 on Build GRCh38
rs144836310 Chr.15: 59619357-59619357 on Build GRCh38
rs138074286 Chr.15: 59619525-59619525 on Build GRCh38
rs763125847 Chr.15: 59619434-59619434 on Build GRCh38

View Full Product Details