Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 38356793-38357018 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CATSPERG
Entrez Gene ID
57828
Gene Name
cation channel sperm associated auxiliary subunit gamma
Gene Aliases
C19orf15
UniGene
Hs.324335
Gene Chromosome Location
Chr.19: 38335563-38370949 on Build GRCh38
Transcripts
NM_021185.4 XM_005259114.4 XM_005259115.4 XM_005259116.2 XM_006723308.3 XM_006723310.2 XM_006723311.2 XM_011527174.2 XM_011527175.2 XM_011527176.2 XM_011527177.2 XM_011527178.2 XM_011527179.1 XM_011527182.2 XM_017027053.1 XM_017027054.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7239677 c.1239G>A p.V413V Chr.19: 38356806-38356806 on Build GRCh38
254585 c.215A>T p.Q72L Chr.19: 38356841-38356841 on Build GRCh38
1750863 c.1274A>T p.Q425L Chr.19: 38356841-38356841 on Build GRCh38
7085834 c.1280T>C p.V427A Chr.19: 38356847-38356847 on Build GRCh38
5612464 c.1264A>T p.S422C Chr.19: 38356810-38356810 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs562520345 Chr.19: 38356815-38356815 on Build GRCh38
rs752998252 Chr.19: 38356882-38356882 on Build GRCh38
rs768411378 Chr.19: 38356829-38356829 on Build GRCh38
rs996256788 Chr.19: 38356881-38356881 on Build GRCh38
rs375444012 Chr.19: 38356867-38356867 on Build GRCh38
rs139287144 Chr.19: 38356835-38356835 on Build GRCh38
rs971597434 Chr.19: 38356991-38356991 on Build GRCh38
rs745819007 Chr.19: 38356812-38356812 on Build GRCh38
rs894932496 Chr.19: 38356891-38356891 on Build GRCh38
rs749176186 Chr.19: 38356821-38356821 on Build GRCh38
rs764276059 Chr.19: 38356884-38356884 on Build GRCh38
rs766805044 Chr.19: 38356978-38356978 on Build GRCh38
rs527289565 Chr.19: 38356937-38356937 on Build GRCh38
rs1027880410 Chr.19: 38356918-38356918 on Build GRCh38
rs549086866 Chr.19: 38356902-38356902 on Build GRCh38
rs763363222 Chr.19: 38356883-38356883 on Build GRCh38
rs368262357 Chr.19: 38356871-38356871 on Build GRCh38
rs551404628 Chr.19: 38356864-38356864 on Build GRCh38
rs751815675 Chr.19: 38356909-38356909 on Build GRCh38
rs888026582 Chr.19: 38356921-38356921 on Build GRCh38
rs1005023688 Chr.19: 38356925-38356925 on Build GRCh38
rs566733516 Chr.19: 38356913-38356913 on Build GRCh38
rs765569963 Chr.19: 38356880-38356880 on Build GRCh38
rs779721512 Chr.19: 38356820-38356820 on Build GRCh38
rs772005025 Chr.19: 38356895-38356895 on Build GRCh38
rs771739027 Chr.19: 38356866-38356866 on Build GRCh38
rs774196898 Chr.19: 38356834-38356834 on Build GRCh38

View Full Product Details