Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161749735-161749974 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP12
Entrez Gene ID
11266
Gene Name
dual specificity phosphatase 12
Gene Aliases
DUSP1, YVH1
UniGene
Hs.416216
Gene Chromosome Location
Chr.1: 161749692-161757164 on Build GRCh38
Transcripts
NM_007240.2 XM_005244862.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6650310 c.120G>T p.A40A Chr.1: 161749921-161749921 on Build GRCh38
1335705 c.132G>A p.E44E Chr.1: 161749933-161749933 on Build GRCh38
6567150 c.34G>A p.E12K Chr.1: 161749835-161749835 on Build GRCh38
1319802 c.93G>C p.Q31H Chr.1: 161749894-161749894 on Build GRCh38
6295547 c.166G>T p.V56L Chr.1: 161749967-161749967 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746240714 Chr.1: 161749788-161749788 on Build GRCh38
rs146495672 Chr.1: 161749946-161749946 on Build GRCh38
rs746185804 Chr.1: 161749854-161749854 on Build GRCh38
rs150594405 Chr.1: 161749847-161749847 on Build GRCh38
rs750902601 Chr.1: 161749951-161749951 on Build GRCh38
rs771949809 Chr.1: 161749831-161749831 on Build GRCh38
rs774257991 Chr.1: 161749924-161749924 on Build GRCh38
rs367765196 Chr.1: 161749937-161749937 on Build GRCh38
rs775580032 Chr.1: 161749846-161749846 on Build GRCh38
rs748264213 Chr.1: 161749905-161749905 on Build GRCh38
rs762157914 Chr.1: 161749859-161749860 on Build GRCh38
rs35106830 Chr.1: 161749953-161749953 on Build GRCh38
rs776838624 Chr.1: 161749801-161749801 on Build GRCh38
rs761009700 Chr.1: 161749796-161749796 on Build GRCh38
rs536036743 Chr.1: 161749757-161749757 on Build GRCh38
rs768589864 Chr.1: 161749800-161749800 on Build GRCh38
rs757634240 Chr.1: 161749852-161749852 on Build GRCh38
rs766323982 Chr.1: 161749823-161749823 on Build GRCh38
rs775714884 Chr.1: 161749795-161749795 on Build GRCh38
rs1044564131 Chr.1: 161749870-161749870 on Build GRCh38
rs752113173 Chr.1: 161749944-161749944 on Build GRCh38
rs747193973 Chr.1: 161749866-161749866 on Build GRCh38
rs754723718 Chr.1: 161749834-161749834 on Build GRCh38
rs144419580 Chr.1: 161749869-161749869 on Build GRCh38
rs539052931 Chr.1: 161749917-161749917 on Build GRCh38
rs149171825 Chr.1: 161749945-161749945 on Build GRCh38
rs773260246 Chr.1: 161749915-161749915 on Build GRCh38
rs772246099 Chr.1: 161749791-161749791 on Build GRCh38
rs138579064 Chr.1: 161749909-161749909 on Build GRCh38
rs397863929 Chr.1: 161749917-161749917 on Build GRCh38
rs779186714 Chr.1: 161749785-161749785 on Build GRCh38
rs1037214103 Chr.1: 161749837-161749837 on Build GRCh38
rs766270697 Chr.1: 161749922-161749922 on Build GRCh38
rs762862670 Chr.1: 161749818-161749818 on Build GRCh38
rs765322308 Chr.1: 161749920-161749961 on Build GRCh38
rs752261197 Chr.1: 161749848-161749848 on Build GRCh38
rs760741271 Chr.1: 161749929-161749929 on Build GRCh38
rs758742789 Chr.1: 161749860-161749860 on Build GRCh38
rs779431540 Chr.1: 161749853-161749853 on Build GRCh38
rs869127078 Chr.1: 161749794-161749794 on Build GRCh38
rs771523657 Chr.1: 161749770-161749770 on Build GRCh38
rs776516231 Chr.1: 161749875-161749875 on Build GRCh38
rs932842410 Chr.1: 161749764-161749764 on Build GRCh38
rs773101189 Chr.1: 161749817-161749817 on Build GRCh38
rs767200899 Chr.1: 161749941-161749941 on Build GRCh38
rs759050136 Chr.1: 161749936-161749936 on Build GRCh38
rs147989847 Chr.1: 161749826-161749826 on Build GRCh38
rs370225521 Chr.1: 161749861-161749861 on Build GRCh38
rs765280035 Chr.1: 161749816-161749816 on Build GRCh38
rs188951859 Chr.1: 161749815-161749815 on Build GRCh38
rs397862197 Chr.1: 161749792-161749792 on Build GRCh38
rs376939090 Chr.1: 161749813-161749813 on Build GRCh38
rs869209328 Chr.1: 161749918-161749918 on Build GRCh38

View Full Product Details