Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 6678361-6678588 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3
Entrez Gene ID
718
Gene Name
complement component 3
Gene Aliases
AHUS5, ARMD9, ASP, C3a, C3b, CPAMD1, HEL-S-62p
UniGene
Hs.529053
Gene Chromosome Location
Chr.19: 6677835-6720682 on Build GRCh38
Transcripts
NM_000064.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5449360 c.4664C>T p.S1555F Chr.19: 6678422-6678422 on Build GRCh38
3893821 c.4686C>T p.I1562I Chr.19: 6678400-6678400 on Build GRCh38
4642392 c.4634A>G p.Y1545C Chr.19: 6678452-6678452 on Build GRCh38
1198768 c.4687A>G p.M1563V Chr.19: 6678399-6678399 on Build GRCh38
5548229 c.4631-5delC p.? Chr.19: 6678460-6678460 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs911659473 Chr.19: 6678505-6678505 on Build GRCh38
rs111595742 Chr.19: 6678457-6678457 on Build GRCh38
rs375264433 Chr.19: 6678502-6678502 on Build GRCh38
rs753610060 Chr.19: 6678499-6678499 on Build GRCh38
rs765397948 Chr.19: 6678481-6678481 on Build GRCh38
rs764593615 Chr.19: 6678392-6678392 on Build GRCh38
rs1044307395 Chr.19: 6678498-6678498 on Build GRCh38
rs924244787 Chr.19: 6678526-6678526 on Build GRCh38
rs752668279 Chr.19: 6678424-6678424 on Build GRCh38
rs1010056052 Chr.19: 6678537-6678537 on Build GRCh38
rs545349985 Chr.19: 6678448-6678448 on Build GRCh38
rs144156850 Chr.19: 6678433-6678433 on Build GRCh38
rs549423358 Chr.19: 6678531-6678531 on Build GRCh38
rs750498103 Chr.19: 6678408-6678408 on Build GRCh38
rs149202905 Chr.19: 6678441-6678441 on Build GRCh38
rs113432208 Chr.19: 6678458-6678458 on Build GRCh38
rs11569564 Chr.19: 6678484-6678484 on Build GRCh38
rs759414386 Chr.19: 6678490-6678491 on Build GRCh38
rs770510549 Chr.19: 6678532-6678532 on Build GRCh38
rs756063740 Chr.19: 6678491-6678491 on Build GRCh38
rs542668902 Chr.19: 6678421-6678421 on Build GRCh38
rs765432507 Chr.19: 6678403-6678403 on Build GRCh38
rs762196772 Chr.19: 6678476-6678476 on Build GRCh38
rs775109096 Chr.19: 6678461-6678461 on Build GRCh38
rs751621240 Chr.19: 6678487-6678487 on Build GRCh38
rs777040887 Chr.19: 6678395-6678395 on Build GRCh38
rs143373043 Chr.19: 6678508-6678508 on Build GRCh38
rs776863964 Chr.19: 6678475-6678475 on Build GRCh38
rs189948635 Chr.19: 6678451-6678451 on Build GRCh38
rs761954547 Chr.19: 6678397-6678397 on Build GRCh38
rs750559710 Chr.19: 6678422-6678422 on Build GRCh38
rs868660644 Chr.19: 6678416-6678416 on Build GRCh38
rs559302635 Chr.19: 6678436-6678436 on Build GRCh38
rs200834388 Chr.19: 6678402-6678402 on Build GRCh38
rs755393196 Chr.19: 6678474-6678474 on Build GRCh38
rs780839437 Chr.19: 6678535-6678535 on Build GRCh38
rs11569565 Chr.19: 6678463-6678463 on Build GRCh38
rs139381845 Chr.19: 6678419-6678419 on Build GRCh38
rs116302413 Chr.19: 6678464-6678464 on Build GRCh38
rs778590435 Chr.19: 6678443-6678443 on Build GRCh38
rs986309144 Chr.19: 6678490-6678490 on Build GRCh38
rs748672252 Chr.19: 6678519-6678519 on Build GRCh38

View Full Product Details