Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 56952043-56952268 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8B
Entrez Gene ID
732
Gene Name
complement component 8, beta polypeptide
Gene Aliases
C82
UniGene
Hs.391835
Gene Chromosome Location
Chr.1: 56929210-56974383 on Build GRCh38
Transcripts
NM_000066.3 NM_001278543.1 NM_001278544.1 XM_017002235.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1246964 c.550_553delAACA p.N184fs*22 Chr.1: 56952161-56952164 on Build GRCh38
7336007 c.534-1G>A p.? Chr.1: 56952181-56952181 on Build GRCh38
6616674 c.578A>C p.H193P Chr.1: 56952136-56952136 on Build GRCh38
4981743 c.559G>C p.E187Q Chr.1: 56952155-56952155 on Build GRCh38
5847061 c.602C>T p.S201F Chr.1: 56952112-56952112 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201159708 Chr.1: 56952222-56952222 on Build GRCh38
rs142822254 Chr.1: 56952109-56952109 on Build GRCh38
rs1049542804 Chr.1: 56952185-56952185 on Build GRCh38
rs375513059 Chr.1: 56952140-56952140 on Build GRCh38
rs201174086 Chr.1: 56952178-56952178 on Build GRCh38
rs371177945 Chr.1: 56952121-56952121 on Build GRCh38
rs530066947 Chr.1: 56952148-56952148 on Build GRCh38
rs398122867 Chr.1: 56952109-56952109 on Build GRCh38
rs777143510 Chr.1: 56952223-56952223 on Build GRCh38
rs543235905 Chr.1: 56952164-56952164 on Build GRCh38
rs904564646 Chr.1: 56952074-56952074 on Build GRCh38
rs148621962 Chr.1: 56952160-56952160 on Build GRCh38
rs374780772 Chr.1: 56952218-56952218 on Build GRCh38
rs747595186 Chr.1: 56952214-56952216 on Build GRCh38
rs758410802 Chr.1: 56952205-56952205 on Build GRCh38
rs202137767 Chr.1: 56952157-56952157 on Build GRCh38
rs752249954 Chr.1: 56952204-56952204 on Build GRCh38
rs753583783 Chr.1: 56952183-56952183 on Build GRCh38
rs143870427 Chr.1: 56952129-56952129 on Build GRCh38
rs576004874 Chr.1: 56952094-56952094 on Build GRCh38
rs34438223 Chr.1: 56952108-56952108 on Build GRCh38
rs137965251 Chr.1: 56952114-56952114 on Build GRCh38
rs560947620 Chr.1: 56952151-56952151 on Build GRCh38
rs1024437964 Chr.1: 56952069-56952069 on Build GRCh38
rs572299334 Chr.1: 56952111-56952111 on Build GRCh38
rs751857773 Chr.1: 56952097-56952097 on Build GRCh38
rs200607505 Chr.1: 56952210-56952210 on Build GRCh38
rs769039585 Chr.1: 56952162-56952162 on Build GRCh38
rs758396641 Chr.1: 56952120-56952120 on Build GRCh38
rs760443732 Chr.1: 56952101-56952101 on Build GRCh38
rs148088012 Chr.1: 56952090-56952090 on Build GRCh38
rs745980920 Chr.1: 56952141-56952141 on Build GRCh38
rs562696863 Chr.1: 56952211-56952211 on Build GRCh38
rs759239969 Chr.1: 56952194-56952194 on Build GRCh38
rs866480176 Chr.1: 56952133-56952133 on Build GRCh38
rs776295488 Chr.1: 56952160-56952161 on Build GRCh38
rs763423680 Chr.1: 56952091-56952091 on Build GRCh38
rs780175316 Chr.1: 56952232-56952232 on Build GRCh38
rs781012145 Chr.1: 56952221-56952221 on Build GRCh38
rs921827461 Chr.1: 56952171-56952171 on Build GRCh38
rs140192392 Chr.1: 56952169-56952169 on Build GRCh38
rs1000158849 Chr.1: 56952084-56952084 on Build GRCh38
rs764897887 Chr.1: 56952195-56952195 on Build GRCh38
rs199704406 Chr.1: 56952219-56952219 on Build GRCh38
rs766080403 Chr.1: 56952182-56952182 on Build GRCh38
rs963090908 Chr.1: 56952193-56952193 on Build GRCh38

View Full Product Details