Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 74037606-74037845 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELN
Entrez Gene ID
2006
Gene Name
elastin
Gene Aliases
SVAS, WBS, WS
UniGene
Hs.647061
Gene Chromosome Location
Chr.7: 74027772-74069907 on Build GRCh38
Transcripts
NM_000501.3 NM_001081752.2 NM_001081753.2 NM_001081754.2 NM_001081755.2 NM_001278912.1 NM_001278913.1 NM_001278914.1 NM_001278915.1 NM_001278916.1 NM_001278917.1 NM_001278918.1 NM_001278939.1 XM_005250187.1 XM_005250188.1 XM_011515868.1 XM_011515869.1 XM_011515870.1 XM_011515871.1 XM_011515872.1 XM_011515873.1 XM_011515874.1 XM_011515875.1 XM_011515876.1 XM_011515877.1 XM_017011813.1 XM_017011814.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6193488 c.169G>C p.G57R Chr.7: 74037712-74037712 on Build GRCh38
5207009 c.178G>C p.G60R Chr.7: 74037721-74037721 on Build GRCh38
6193489 c.169G>C p.G57R Chr.7: 74037712-74037712 on Build GRCh38
3367021 c.165G>A p.A55A Chr.7: 74037708-74037708 on Build GRCh38
3640413 c.168G>T p.L56L Chr.7: 74037711-74037711 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782562859 Chr.7: 74037627-74037627 on Build GRCh38
rs782141308 Chr.7: 74037719-74037719 on Build GRCh38
rs896537913 Chr.7: 74037679-74037679 on Build GRCh38
rs782475698 Chr.7: 74037749-74037749 on Build GRCh38
rs782406469 Chr.7: 74037681-74037681 on Build GRCh38
rs141089547 Chr.7: 74037775-74037775 on Build GRCh38
rs781790610 Chr.7: 74037735-74037735 on Build GRCh38
rs782191688 Chr.7: 74037634-74037634 on Build GRCh38
rs2301995 Chr.7: 74037810-74037810 on Build GRCh38
rs781847678 Chr.7: 74037774-74037774 on Build GRCh38
rs1030566999 Chr.7: 74037781-74037781 on Build GRCh38
rs782359717 Chr.7: 74037673-74037673 on Build GRCh38
rs371258256 Chr.7: 74037779-74037779 on Build GRCh38
rs782628350 Chr.7: 74037677-74037677 on Build GRCh38
rs377019817 Chr.7: 74037812-74037812 on Build GRCh38
rs376512299 Chr.7: 74037708-74037708 on Build GRCh38
rs782528515 Chr.7: 74037689-74037689 on Build GRCh38
rs781859547 Chr.7: 74037756-74037756 on Build GRCh38
rs782217420 Chr.7: 74037659-74037659 on Build GRCh38
rs370961040 Chr.7: 74037752-74037752 on Build GRCh38
rs782238532 Chr.7: 74037794-74037794 on Build GRCh38
rs540977629 Chr.7: 74037652-74037652 on Build GRCh38
rs781867378 Chr.7: 74037714-74037714 on Build GRCh38
rs782096458 Chr.7: 74037705-74037705 on Build GRCh38
rs374252054 Chr.7: 74037687-74037687 on Build GRCh38
rs374473857 Chr.7: 74037757-74037757 on Build GRCh38
rs147723448 Chr.7: 74037763-74037763 on Build GRCh38
rs782208197 Chr.7: 74037778-74037778 on Build GRCh38
rs781943228 Chr.7: 74037693-74037693 on Build GRCh38
rs782264874 Chr.7: 74037680-74037680 on Build GRCh38
rs782319280 Chr.7: 74037691-74037691 on Build GRCh38

View Full Product Details