Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140558578-140558849 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APBB3
Entrez Gene ID
10307
Gene Name
amyloid beta precursor protein binding family B member 3
Gene Aliases
FE65L2, SRA
UniGene
Hs.529449
Gene Chromosome Location
Chr.5: 140558268-140564604 on Build GRCh38
Transcripts
NM_006051.3 NM_133172.2 NM_133173.2 NM_133174.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5060662 c.1310G>A p.R437H Chr.5: 140558757-140558757 on Build GRCh38
1433632 c.1274C>A p.S425Y Chr.5: 140558793-140558793 on Build GRCh38
1433631 c.1305G>T p.Q435H Chr.5: 140558762-140558762 on Build GRCh38
4524885 c.1298G>A p.G433D Chr.5: 140558769-140558769 on Build GRCh38
7000060 c.1379T>C p.L460P Chr.5: 140558688-140558688 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745339001 Chr.5: 140558766-140558766 on Build GRCh38
rs745441682 Chr.5: 140558705-140558705 on Build GRCh38
rs751180124 Chr.5: 140558627-140558631 on Build GRCh38
rs746337320 Chr.5: 140558745-140558745 on Build GRCh38
rs34926962 Chr.5: 140558785-140558785 on Build GRCh38
rs536302777 Chr.5: 140558809-140558809 on Build GRCh38
rs748565423 Chr.5: 140558699-140558699 on Build GRCh38
rs199704720 Chr.5: 140558784-140558784 on Build GRCh38
rs756002552 Chr.5: 140558781-140558782 on Build GRCh38
rs778410690 Chr.5: 140558763-140558763 on Build GRCh38
rs143491686 Chr.5: 140558657-140558657 on Build GRCh38
rs374702672 Chr.5: 140558653-140558653 on Build GRCh38
rs148042615 Chr.5: 140558703-140558703 on Build GRCh38
rs370414260 Chr.5: 140558641-140558641 on Build GRCh38
rs776227488 Chr.5: 140558797-140558797 on Build GRCh38
rs768520917 Chr.5: 140558729-140558729 on Build GRCh38
rs770340112 Chr.5: 140558700-140558700 on Build GRCh38
rs748777523 Chr.5: 140558638-140558638 on Build GRCh38
rs752462282 Chr.5: 140558647-140558647 on Build GRCh38
rs146302480 Chr.5: 140558759-140558759 on Build GRCh38
rs148938296 Chr.5: 140558718-140558718 on Build GRCh38
rs764090820 Chr.5: 140558656-140558656 on Build GRCh38
rs765187519 Chr.5: 140558667-140558667 on Build GRCh38
rs757074030 Chr.5: 140558630-140558630 on Build GRCh38
rs781471610 Chr.5: 140558758-140558758 on Build GRCh38
rs961651106 Chr.5: 140558682-140558682 on Build GRCh38
rs994087660 Chr.5: 140558616-140558616 on Build GRCh38
rs564829716 Chr.5: 140558622-140558622 on Build GRCh38
rs773944900 Chr.5: 140558646-140558646 on Build GRCh38
rs761829691 Chr.5: 140558664-140558664 on Build GRCh38
rs761036999 Chr.5: 140558683-140558685 on Build GRCh38
rs111366985 Chr.5: 140558780-140558780 on Build GRCh38
rs745771417 Chr.5: 140558722-140558723 on Build GRCh38
rs761721268 Chr.5: 140558806-140558806 on Build GRCh38
rs529722873 Chr.5: 140558671-140558671 on Build GRCh38
rs760522044 Chr.5: 140558725-140558725 on Build GRCh38
rs537032916 Chr.5: 140558736-140558736 on Build GRCh38
rs143692635 Chr.5: 140558752-140558752 on Build GRCh38
rs377738702 Chr.5: 140558672-140558672 on Build GRCh38
rs755227669 Chr.5: 140558757-140558757 on Build GRCh38
rs532222938 Chr.5: 140558624-140558624 on Build GRCh38
rs760613759 Chr.5: 140558655-140558655 on Build GRCh38
rs548169457 Chr.5: 140558692-140558692 on Build GRCh38
rs150554513 Chr.5: 140558737-140558737 on Build GRCh38
rs781561764 Chr.5: 140558698-140558698 on Build GRCh38
rs771757069 Chr.5: 140558768-140558768 on Build GRCh38
rs17855437 Chr.5: 140558632-140558632 on Build GRCh38
rs778229844 Chr.5: 140558701-140558701 on Build GRCh38
rs753336111 Chr.5: 140558695-140558695 on Build GRCh38
rs765626152 Chr.5: 140558642-140558642 on Build GRCh38
rs145698446 Chr.5: 140558652-140558652 on Build GRCh38
rs138766975 Chr.5: 140558751-140558751 on Build GRCh38
rs746617921 Chr.5: 140558782-140558782 on Build GRCh38
rs773253585 Chr.5: 140558812-140558812 on Build GRCh38
rs138614728 Chr.5: 140558617-140558617 on Build GRCh38
rs868116147 Chr.5: 140558670-140558670 on Build GRCh38
rs780325641 Chr.5: 140558687-140558687 on Build GRCh38
rs758829612 Chr.5: 140558746-140558746 on Build GRCh38

View Full Product Details