Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32211614-32211877 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCDC2B
Entrez Gene ID
149069
Gene Name
doublecortin domain containing 2B
Gene Aliases
-
UniGene
Hs.424588
Gene Chromosome Location
Chr.1: 32208813-32216196 on Build GRCh38
Transcripts
NM_001099434.1 XM_017000413.1 XM_017000414.1 XM_011541795.2 XM_017001814.1 XM_017001815.1 XM_017001817.1 XM_017001818.1 XM_017001819.1 XM_017001821.1 XM_017001827.1 XM_017001828.1

Gene Symbol
TMEM234
Entrez Gene ID
56063
Gene Name
transmembrane protein 234
Gene Aliases
AASL548, C1orf91, PRO1105, RP4-622L5, dJ622L5.7
UniGene
Hs.272299
Gene Chromosome Location
Chr.1: 32204807-32222371 on Build GRCh38
Transcripts
NM_001099434.1 XM_017000413.1 XM_017000414.1 XM_011541795.2 XM_017001814.1 XM_017001815.1 XM_017001817.1 XM_017001818.1 XM_017001819.1 XM_017001821.1 XM_017001827.1 XM_017001828.1

SNP Detail

SNP ID Chromosome Location
rs963109835 Chr.1: 32211675-32211675 on Build GRCh38
rs986216846 Chr.1: 32211681-32211681 on Build GRCh38
rs747383037 Chr.1: 32211856-32211856 on Build GRCh38
rs759489138 Chr.1: 32211815-32211815 on Build GRCh38
rs760485939 Chr.1: 32211838-32211839 on Build GRCh38
rs777863878 Chr.1: 32211750-32211750 on Build GRCh38
rs565670123 Chr.1: 32211674-32211674 on Build GRCh38
rs747473535 Chr.1: 32211786-32211786 on Build GRCh38
rs747612916 Chr.1: 32211799-32211799 on Build GRCh38
rs71646365 Chr.1: 32211657-32211657 on Build GRCh38
rs536393129 Chr.1: 32211816-32211816 on Build GRCh38
rs368080232 Chr.1: 32211798-32211798 on Build GRCh38
rs182464428 Chr.1: 32211769-32211769 on Build GRCh38
rs901608327 Chr.1: 32211834-32211834 on Build GRCh38
rs772316436 Chr.1: 32211731-32211731 on Build GRCh38
rs967930738 Chr.1: 32211655-32211655 on Build GRCh38
rs929786680 Chr.1: 32211773-32211773 on Build GRCh38
rs202211725 Chr.1: 32211776-32211776 on Build GRCh38
rs750682144 Chr.1: 32211845-32211845 on Build GRCh38
rs766240063 Chr.1: 32211811-32211811 on Build GRCh38
rs780347925 Chr.1: 32211849-32211849 on Build GRCh38
rs200489306 Chr.1: 32211819-32211819 on Build GRCh38
rs765473851 Chr.1: 32211843-32211843 on Build GRCh38
rs534880580 Chr.1: 32211727-32211727 on Build GRCh38
rs751590172 Chr.1: 32211722-32211722 on Build GRCh38
rs753877276 Chr.1: 32211761-32211761 on Build GRCh38
rs1020551344 Chr.1: 32211649-32211649 on Build GRCh38
rs972830570 Chr.1: 32211694-32211694 on Build GRCh38
rs115145784 Chr.1: 32211803-32211803 on Build GRCh38
rs903466415 Chr.1: 32211854-32211854 on Build GRCh38
rs953481570 Chr.1: 32211678-32211678 on Build GRCh38
rs757341336 Chr.1: 32211829-32211829 on Build GRCh38
rs368115283 Chr.1: 32211647-32211647 on Build GRCh38
rs147072705 Chr.1: 32211804-32211804 on Build GRCh38
rs538049148 Chr.1: 32211719-32211719 on Build GRCh38
rs762814113 Chr.1: 32211808-32211808 on Build GRCh38
rs746149653 Chr.1: 32211748-32211748 on Build GRCh38
rs769066118 Chr.1: 32211790-32211790 on Build GRCh38
rs199902105 Chr.1: 32211770-32211770 on Build GRCh38
rs780505681 Chr.1: 32211777-32211777 on Build GRCh38

View Full Product Details