Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 972027-972298 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARID3A
Entrez Gene ID
1820
Gene Name
AT-rich interaction domain 3A
Gene Aliases
BRIGHT, DRIL1, DRIL3, E2FBP1
UniGene
Hs.501296
Gene Chromosome Location
Chr.19: 925733-975934 on Build GRCh38
Transcripts
NM_005224.2 XM_005259513.4 XM_005259514.4 XM_017026445.1 XM_017026446.1 XM_017026447.1

SNP Detail

SNP ID Chromosome Location
rs753037429 Chr.19: 972067-972067 on Build GRCh38
rs397773771 Chr.19: 972247-972248 on Build GRCh38
rs908234480 Chr.19: 972157-972157 on Build GRCh38
rs552277698 Chr.19: 972105-972105 on Build GRCh38
rs112390252 Chr.19: 972151-972151 on Build GRCh38
rs774105029 Chr.19: 972053-972053 on Build GRCh38
rs904507995 Chr.19: 972241-972248 on Build GRCh38
rs71948956 Chr.19: 972232-972233 on Build GRCh38
rs976393536 Chr.19: 972161-972161 on Build GRCh38
rs921519448 Chr.19: 972164-972164 on Build GRCh38
rs557455807 Chr.19: 972224-972224 on Build GRCh38
rs1010712432 Chr.19: 972237-972248 on Build GRCh38
rs754041528 Chr.19: 972270-972270 on Build GRCh38
rs796075096 Chr.19: 972247-972248 on Build GRCh38
rs367651447 Chr.19: 972152-972152 on Build GRCh38
rs549835384 Chr.19: 972124-972124 on Build GRCh38
rs559335883 Chr.19: 972056-972056 on Build GRCh38
rs372795322 Chr.19: 972068-972068 on Build GRCh38
rs991945415 Chr.19: 972102-972102 on Build GRCh38
rs1012711103 Chr.19: 972137-972137 on Build GRCh38
rs768306091 Chr.19: 972051-972051 on Build GRCh38
rs1053993963 Chr.19: 972187-972187 on Build GRCh38
rs960230996 Chr.19: 972246-972247 on Build GRCh38
rs933145976 Chr.19: 972223-972232 on Build GRCh38
rs10637085 Chr.19: 972222-972223 on Build GRCh38
rs557861692 Chr.19: 972201-972201 on Build GRCh38
rs540168766 Chr.19: 972190-972190 on Build GRCh38
rs1003923059 Chr.19: 972117-972117 on Build GRCh38
rs550688908 Chr.19: 972129-972129 on Build GRCh38
rs185134878 Chr.19: 972266-972266 on Build GRCh38
rs777975933 Chr.19: 972083-972083 on Build GRCh38
rs761223505 Chr.19: 972237-972238 on Build GRCh38
rs1043073617 Chr.19: 972240-972241 on Build GRCh38
rs570858435 Chr.19: 972106-972106 on Build GRCh38
rs894431359 Chr.19: 972250-972250 on Build GRCh38
rs201619021 Chr.19: 972100-972100 on Build GRCh38
rs891879895 Chr.19: 972234-972251 on Build GRCh38
rs760137708 Chr.19: 972057-972057 on Build GRCh38
rs965087213 Chr.19: 972160-972160 on Build GRCh38
rs945480016 Chr.19: 972202-972202 on Build GRCh38
rs1003500144 Chr.19: 972246-972246 on Build GRCh38
rs757462936 Chr.19: 972090-972090 on Build GRCh38
rs1027257443 Chr.19: 972269-972269 on Build GRCh38
rs1034544904 Chr.19: 972097-972097 on Build GRCh38
rs906586472 Chr.19: 972226-972226 on Build GRCh38
rs781252359 Chr.19: 972098-972098 on Build GRCh38
rs958576042 Chr.19: 972130-972130 on Build GRCh38
rs537930704 Chr.19: 972122-972122 on Build GRCh38
rs761544461 Chr.19: 972054-972054 on Build GRCh38
rs751833745 Chr.19: 972084-972084 on Build GRCh38
rs921792694 Chr.19: 972163-972163 on Build GRCh38
rs538442292 Chr.19: 972153-972153 on Build GRCh38
rs1026195680 Chr.19: 972251-972252 on Build GRCh38
rs767204634 Chr.19: 972055-972055 on Build GRCh38
rs1548666 Chr.19: 972248-972248 on Build GRCh38
rs566242422 Chr.19: 972179-972179 on Build GRCh38
rs1001807257 Chr.19: 972245-972248 on Build GRCh38
rs765729766 Chr.19: 972060-972060 on Build GRCh38

View Full Product Details