Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 138788245-138788494 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC28A
Entrez Gene ID
25901
Gene Name
coiled-coil domain containing 28A
Gene Aliases
C6orf80, CCRL1AP
UniGene
Hs.412019
Gene Chromosome Location
Chr.6: 138773517-138793319 on Build GRCh38
Transcripts
NM_015439.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5884313 c.751G>T p.E251* Chr.6: 138788369-138788369 on Build GRCh38
1440815 c.754G>T p.E252* Chr.6: 138788372-138788372 on Build GRCh38
3621116 c.767C>T p.S256F Chr.6: 138788385-138788385 on Build GRCh38
6403305 c.758T>G p.L253W Chr.6: 138788376-138788376 on Build GRCh38
345807 c.767C>A p.S256Y Chr.6: 138788385-138788385 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781460692 Chr.6: 138788348-138788348 on Build GRCh38
rs765500495 Chr.6: 138788452-138788452 on Build GRCh38
rs751800638 Chr.6: 138788426-138788426 on Build GRCh38
rs777704295 Chr.6: 138788440-138788440 on Build GRCh38
rs190507166 Chr.6: 138788423-138788423 on Build GRCh38
rs1011451621 Chr.6: 138788420-138788420 on Build GRCh38
rs759222705 Chr.6: 138788389-138788389 on Build GRCh38
rs373271860 Chr.6: 138788435-138788435 on Build GRCh38
rs745460530 Chr.6: 138788346-138788347 on Build GRCh38
rs939385457 Chr.6: 138788456-138788456 on Build GRCh38
rs748075336 Chr.6: 138788362-138788362 on Build GRCh38
rs769200535 Chr.6: 138788297-138788297 on Build GRCh38
rs566921393 Chr.6: 138788432-138788432 on Build GRCh38
rs772875496 Chr.6: 138788363-138788363 on Build GRCh38
rs199859235 Chr.6: 138788424-138788424 on Build GRCh38
rs766987304 Chr.6: 138788391-138788391 on Build GRCh38
rs372508880 Chr.6: 138788399-138788399 on Build GRCh38
rs773939394 Chr.6: 138788387-138788387 on Build GRCh38
rs749042682 Chr.6: 138788365-138788365 on Build GRCh38
rs755300304 Chr.6: 138788431-138788431 on Build GRCh38
rs756149308 Chr.6: 138788434-138788434 on Build GRCh38
rs909903710 Chr.6: 138788268-138788268 on Build GRCh38
rs889025047 Chr.6: 138788402-138788402 on Build GRCh38
rs201106266 Chr.6: 138788385-138788385 on Build GRCh38
rs761981577 Chr.6: 138788354-138788354 on Build GRCh38
rs775033630 Chr.6: 138788392-138788392 on Build GRCh38
rs765842184 Chr.6: 138788422-138788422 on Build GRCh38

View Full Product Details