Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 69260964-69261223 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLCE
Entrez Gene ID
26035
Gene Name
glucuronic acid epimerase
Gene Aliases
HSEPI
UniGene
Hs.183006
Gene Chromosome Location
Chr.15: 69160555-69272209 on Build GRCh38
Transcripts
NM_001324091.1 NM_001324092.1 NM_001324093.1 NM_001324094.1 NM_015554.2 XM_005254298.3 XM_017022073.1 XM_017022074.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1478312 c.587G>T p.G196V Chr.15: 69261087-69261087 on Build GRCh38
3711845 c.700G>A p.E234K Chr.15: 69261200-69261200 on Build GRCh38
4556983 c.709G>C p.E237Q Chr.15: 69261209-69261209 on Build GRCh38
1608627 c.589G>A p.V197M Chr.15: 69261089-69261089 on Build GRCh38
3734524 c.715G>T p.A239S Chr.15: 69261215-69261215 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747442490 Chr.15: 69261061-69261061 on Build GRCh38
rs560361543 Chr.15: 69261083-69261083 on Build GRCh38
rs755691693 Chr.15: 69261037-69261037 on Build GRCh38
rs913562650 Chr.15: 69261001-69261001 on Build GRCh38
rs374101492 Chr.15: 69261076-69261076 on Build GRCh38
rs767390684 Chr.15: 69261187-69261187 on Build GRCh38
rs146050579 Chr.15: 69261102-69261102 on Build GRCh38
rs536473268 Chr.15: 69261021-69261021 on Build GRCh38
rs372095938 Chr.15: 69261044-69261044 on Build GRCh38
rs758922576 Chr.15: 69261053-69261053 on Build GRCh38
rs780475023 Chr.15: 69261057-69261057 on Build GRCh38
rs537655899 Chr.15: 69261132-69261132 on Build GRCh38
rs759454453 Chr.15: 69261178-69261178 on Build GRCh38
rs909244342 Chr.15: 69261192-69261192 on Build GRCh38
rs748787270 Chr.15: 69261046-69261046 on Build GRCh38
rs997659829 Chr.15: 69261088-69261088 on Build GRCh38
rs369532089 Chr.15: 69261065-69261065 on Build GRCh38
rs760766363 Chr.15: 69261010-69261010 on Build GRCh38
rs774419888 Chr.15: 69261168-69261168 on Build GRCh38
rs199691243 Chr.15: 69261094-69261094 on Build GRCh38
rs770994860 Chr.15: 69261167-69261167 on Build GRCh38
rs928814693 Chr.15: 69261058-69261058 on Build GRCh38
rs375630252 Chr.15: 69261062-69261062 on Build GRCh38
rs777372583 Chr.15: 69261111-69261111 on Build GRCh38
rs112026521 Chr.15: 69261035-69261035 on Build GRCh38
rs755724127 Chr.15: 69261078-69261078 on Build GRCh38
rs34316690 Chr.15: 69261016-69261017 on Build GRCh38
rs986392537 Chr.15: 69261115-69261115 on Build GRCh38

View Full Product Details