Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 19661048-19661315 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH3A2
Entrez Gene ID
224
Gene Name
aldehyde dehydrogenase 3 family member A2
Gene Aliases
ALDH10, FALDH, SLS
UniGene
Hs.499886
Gene Chromosome Location
Chr.17: 19648136-19677596 on Build GRCh38
Transcripts
NM_000382.2 NM_001031806.1 XM_011523732.1 XM_011523733.1 XM_017024355.1 XM_017024356.1 XM_017024357.1 XM_017024358.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6043054 c.932G>A p.R311H Chr.17: 19661260-19661260 on Build GRCh38
7400110 c.937A>T p.I313L Chr.17: 19661265-19661265 on Build GRCh38
6598272 c.897G>T p.K299N Chr.17: 19661225-19661225 on Build GRCh38
7189815 c.824A>G p.E275G Chr.17: 19661152-19661152 on Build GRCh38
1679644 c.872T>A p.I291K Chr.17: 19661200-19661200 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs387906255 Chr.17: 19661137-19661137 on Build GRCh38
rs750698259 Chr.17: 19661211-19661211 on Build GRCh38
rs751079690 Chr.17: 19661241-19661241 on Build GRCh38
rs113625085 Chr.17: 19661077-19661077 on Build GRCh38
rs185343765 Chr.17: 19661098-19661098 on Build GRCh38
rs761003121 Chr.17: 19661212-19661212 on Build GRCh38
rs201985073 Chr.17: 19661134-19661134 on Build GRCh38
rs149453193 Chr.17: 19661202-19661202 on Build GRCh38
rs755458255 Chr.17: 19661237-19661237 on Build GRCh38
rs534244558 Chr.17: 19661259-19661259 on Build GRCh38
rs745558212 Chr.17: 19661247-19661247 on Build GRCh38
rs762044215 Chr.17: 19661080-19661080 on Build GRCh38
rs934749419 Chr.17: 19661160-19661160 on Build GRCh38
rs928378092 Chr.17: 19661079-19661079 on Build GRCh38
rs757812222 Chr.17: 19661111-19661111 on Build GRCh38
rs981134776 Chr.17: 19661071-19661071 on Build GRCh38
rs750718862 Chr.17: 19661088-19661088 on Build GRCh38
rs754226801 Chr.17: 19661231-19661231 on Build GRCh38
rs774221570 Chr.17: 19661279-19661279 on Build GRCh38
rs780521810 Chr.17: 19661101-19661101 on Build GRCh38
rs886052689 Chr.17: 19661246-19661246 on Build GRCh38
rs375078986 Chr.17: 19661284-19661284 on Build GRCh38
rs754390132 Chr.17: 19661104-19661104 on Build GRCh38
rs766766826 Chr.17: 19661217-19661217 on Build GRCh38
rs371157031 Chr.17: 19661137-19661137 on Build GRCh38
rs779945294 Chr.17: 19661260-19661260 on Build GRCh38
rs370799289 Chr.17: 19661264-19661264 on Build GRCh38
rs112971336 Chr.17: 19661112-19661112 on Build GRCh38
rs767647102 Chr.17: 19661082-19661082 on Build GRCh38
rs992796190 Chr.17: 19661177-19661177 on Build GRCh38
rs775538629 Chr.17: 19661173-19661173 on Build GRCh38
rs773389475 Chr.17: 19661292-19661292 on Build GRCh38
rs746519259 Chr.17: 19661121-19661121 on Build GRCh38
rs376353294 Chr.17: 19661244-19661244 on Build GRCh38
rs777290506 Chr.17: 19661114-19661114 on Build GRCh38
rs748243317 Chr.17: 19661283-19661283 on Build GRCh38
rs761803237 Chr.17: 19661198-19661198 on Build GRCh38
rs778832796 Chr.17: 19661188-19661189 on Build GRCh38
rs932658083 Chr.17: 19661187-19661187 on Build GRCh38
rs769763084 Chr.17: 19661145-19661145 on Build GRCh38
rs745645947 Chr.17: 19661139-19661139 on Build GRCh38
rs786204759 Chr.17: 19661229-19661231 on Build GRCh38
rs750697040 Chr.17: 19661229-19661229 on Build GRCh38
rs1057516788 Chr.17: 19661152-19661153 on Build GRCh38
rs150614683 Chr.17: 19661184-19661184 on Build GRCh38
rs779297363 Chr.17: 19661238-19661238 on Build GRCh38
rs762906652 Chr.17: 19661175-19661175 on Build GRCh38
rs774431593 Chr.17: 19661185-19661185 on Build GRCh38
rs756776261 Chr.17: 19661243-19661243 on Build GRCh38
rs72547570 Chr.17: 19661163-19661163 on Build GRCh38
rs751620402 Chr.17: 19661076-19661076 on Build GRCh38

View Full Product Details