Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 57231172-57231397 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AURKC
Entrez Gene ID
6795
Gene Name
aurora kinase C
Gene Aliases
AIE2, AIK3, ARK3, AurC, HEL-S-90, SPGF5, STK13, aurora-C
UniGene
Hs.98338
Gene Chromosome Location
Chr.19: 57230317-57235550 on Build GRCh38
Transcripts
NM_001015878.1 NM_001015879.1 NM_003160.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4632817 c.49G>T p.G17C Chr.19: 57231297-57231297 on Build GRCh38
4632244 c.11C>A p.P4H Chr.19: 57231259-57231259 on Build GRCh38
4961466 c.37G>A p.A13T Chr.19: 57231285-57231285 on Build GRCh38
6416734 c.43C>A p.P15T Chr.19: 57231291-57231291 on Build GRCh38
4825911 c.58+3G>A p.? Chr.19: 57231309-57231309 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs546617287 Chr.19: 57231225-57231225 on Build GRCh38
rs141765733 Chr.19: 57231261-57231261 on Build GRCh38
rs746303923 Chr.19: 57231353-57231353 on Build GRCh38
rs777213018 Chr.19: 57231300-57231300 on Build GRCh38
rs774744694 Chr.19: 57231347-57231347 on Build GRCh38
rs774746946 Chr.19: 57231248-57231248 on Build GRCh38
rs756871158 Chr.19: 57231333-57231333 on Build GRCh38
rs971571344 Chr.19: 57231249-57231249 on Build GRCh38
rs553335951 Chr.19: 57231259-57231259 on Build GRCh38
rs1052325167 Chr.19: 57231229-57231229 on Build GRCh38
rs975373769 Chr.19: 57231290-57231290 on Build GRCh38
rs376021169 Chr.19: 57231373-57231373 on Build GRCh38
rs375232125 Chr.19: 57231299-57231299 on Build GRCh38
rs972317809 Chr.19: 57231346-57231346 on Build GRCh38
rs73633650 Chr.19: 57231260-57231260 on Build GRCh38
rs770034685 Chr.19: 57231372-57231372 on Build GRCh38
rs1016637080 Chr.19: 57231210-57231210 on Build GRCh38
rs568732755 Chr.19: 57231235-57231235 on Build GRCh38
rs182340558 Chr.19: 57231294-57231294 on Build GRCh38
rs1004285802 Chr.19: 57231204-57231204 on Build GRCh38
rs763176940 Chr.19: 57231223-57231223 on Build GRCh38
rs765267337 Chr.19: 57231280-57231280 on Build GRCh38
rs933833181 Chr.19: 57231317-57231317 on Build GRCh38
rs770401705 Chr.19: 57231211-57231211 on Build GRCh38
rs754714440 Chr.19: 57231262-57231262 on Build GRCh38
rs548550020 Chr.19: 57231212-57231212 on Build GRCh38
rs369192834 Chr.19: 57231332-57231332 on Build GRCh38
rs779419217 Chr.19: 57231254-57231255 on Build GRCh38
rs898180023 Chr.19: 57231365-57231365 on Build GRCh38
rs1023200714 Chr.19: 57231245-57231245 on Build GRCh38
rs1038513219 Chr.19: 57231360-57231360 on Build GRCh38
rs748701943 Chr.19: 57231355-57231356 on Build GRCh38
rs969884455 Chr.19: 57231279-57231279 on Build GRCh38
rs533590946 Chr.19: 57231237-57231237 on Build GRCh38
rs774957925 Chr.19: 57231366-57231367 on Build GRCh38
rs914607463 Chr.19: 57231275-57231275 on Build GRCh38
rs372070124 Chr.19: 57231227-57231227 on Build GRCh38
rs990631555 Chr.19: 57231274-57231274 on Build GRCh38
rs758097 Chr.19: 57231335-57231335 on Build GRCh38

View Full Product Details