Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 122560680-122560939 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMBT1
Entrez Gene ID
1755
Gene Name
deleted in malignant brain tumors 1
Gene Aliases
GP340, SAG, muclin
UniGene
Hs.279611
Gene Chromosome Location
Chr.10: 122560665-122643736 on Build GRCh38
Transcripts
NM_001320644.1 NM_004406.2 NM_007329.2 NM_017579.2 XM_006717660.3 XM_006717665.3 XM_011539388.2 XM_011539389.2 XM_011539390.2 XM_011539391.2 XM_011539392.2 XM_011539393.2 XM_011539394.2 XM_011539395.2 XM_011539396.2 XM_011539398.2 XM_011539399.2 XM_011539400.2 XM_011539401.2 XM_011539402.2 XM_011539403.2 XM_011539405.2 XM_011539407.2 XM_011539408.2 XM_011539409.2 XM_011539410.2 XM_011539411.2 XM_011539413.2 XM_011539414.2 XM_011539415.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5370229 c.61+2T>C p.? Chr.10: 122560833-122560833 on Build GRCh38
5370230 c.61+2T>C p.? Chr.10: 122560833-122560833 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs767455945 Chr.10: 122560865-122560865 on Build GRCh38
rs79078159 Chr.10: 122560839-122560839 on Build GRCh38
rs753332164 Chr.10: 122560751-122560751 on Build GRCh38
rs748630395 Chr.10: 122560767-122560767 on Build GRCh38
rs36022775 Chr.10: 122560844-122560845 on Build GRCh38
rs772011371 Chr.10: 122560897-122560899 on Build GRCh38
rs189973248 Chr.10: 122560796-122560796 on Build GRCh38
rs765832719 Chr.10: 122560747-122560747 on Build GRCh38
rs758799949 Chr.10: 122560770-122560770 on Build GRCh38
rs1006568746 Chr.10: 122560720-122560720 on Build GRCh38
rs574591699 Chr.10: 122560706-122560706 on Build GRCh38
rs150642921 Chr.10: 122560785-122560785 on Build GRCh38
rs182661047 Chr.10: 122560838-122560838 on Build GRCh38
rs866585604 Chr.10: 122560773-122560773 on Build GRCh38
rs921251218 Chr.10: 122560886-122560886 on Build GRCh38
rs761814232 Chr.10: 122560850-122560850 on Build GRCh38
rs948220327 Chr.10: 122560905-122560905 on Build GRCh38
rs1023024509 Chr.10: 122560733-122560741 on Build GRCh38
rs1035235599 Chr.10: 122560754-122560754 on Build GRCh38
rs778071942 Chr.10: 122560782-122560782 on Build GRCh38
rs746025070 Chr.10: 122560823-122560823 on Build GRCh38
rs377205455 Chr.10: 122560722-122560722 on Build GRCh38
rs567473612 Chr.10: 122560866-122560866 on Build GRCh38
rs889649027 Chr.10: 122560705-122560705 on Build GRCh38
rs995152341 Chr.10: 122560728-122560728 on Build GRCh38
rs199949820 Chr.10: 122560871-122560871 on Build GRCh38
rs565256131 Chr.10: 122560816-122560816 on Build GRCh38
rs369315785 Chr.10: 122560795-122560795 on Build GRCh38
rs769329857 Chr.10: 122560732-122560732 on Build GRCh38
rs779542621 Chr.10: 122560764-122560764 on Build GRCh38
rs1019322715 Chr.10: 122560759-122560759 on Build GRCh38
rs905475653 Chr.10: 122560736-122560736 on Build GRCh38
rs75610148 Chr.10: 122560784-122560784 on Build GRCh38
rs745575710 Chr.10: 122560788-122560788 on Build GRCh38
rs552127186 Chr.10: 122560802-122560802 on Build GRCh38
rs755446978 Chr.10: 122560752-122560752 on Build GRCh38
rs368735301 Chr.10: 122560762-122560762 on Build GRCh38

View Full Product Details