Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 40555420-40555647 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCR7
Entrez Gene ID
1236
Gene Name
C-C motif chemokine receptor 7
Gene Aliases
BLR2, CC-CKR-7, CCR-7, CD197, CDw197, CMKBR7, EBI1
UniGene
Hs.370036
Gene Chromosome Location
Chr.17: 40553769-40565484 on Build GRCh38
Transcripts
NM_001301714.1 NM_001301716.1 NM_001301717.1 NM_001301718.1 NM_001838.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4151606 c.318C>A p.L106L Chr.17: 40555561-40555561 on Build GRCh38
3517064 c.276G>A p.M92I Chr.17: 40555603-40555603 on Build GRCh38
5055451 c.381C>T p.H127H Chr.17: 40555498-40555498 on Build GRCh38
3517063 c.374G>A p.G125D Chr.17: 40555505-40555505 on Build GRCh38
4670295 c.430A>G p.M144V Chr.17: 40555449-40555449 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766444905 Chr.17: 40555577-40555577 on Build GRCh38
rs746470132 Chr.17: 40555519-40555519 on Build GRCh38
rs867209804 Chr.17: 40555560-40555560 on Build GRCh38
rs187022118 Chr.17: 40555563-40555563 on Build GRCh38
rs962555752 Chr.17: 40555606-40555606 on Build GRCh38
rs770522078 Chr.17: 40555523-40555523 on Build GRCh38
rs891813036 Chr.17: 40555502-40555502 on Build GRCh38
rs753865062 Chr.17: 40555599-40555599 on Build GRCh38
rs778319282 Chr.17: 40555516-40555516 on Build GRCh38
rs755131953 Chr.17: 40555600-40555600 on Build GRCh38
rs536242338 Chr.17: 40555536-40555536 on Build GRCh38
rs374509706 Chr.17: 40555449-40555449 on Build GRCh38
rs765943208 Chr.17: 40555503-40555503 on Build GRCh38
rs369179451 Chr.17: 40555528-40555528 on Build GRCh38
rs766491522 Chr.17: 40555480-40555480 on Build GRCh38
rs372058135 Chr.17: 40555615-40555615 on Build GRCh38
rs371166143 Chr.17: 40555507-40555507 on Build GRCh38
rs755221751 Chr.17: 40555492-40555492 on Build GRCh38
rs748522049 Chr.17: 40555506-40555506 on Build GRCh38
rs764096669 Chr.17: 40555543-40555543 on Build GRCh38
rs147082234 Chr.17: 40555527-40555527 on Build GRCh38
rs756261392 Chr.17: 40555476-40555476 on Build GRCh38
rs761281929 Chr.17: 40555526-40555526 on Build GRCh38
rs754045581 Chr.17: 40555482-40555482 on Build GRCh38
rs750465836 Chr.17: 40555462-40555462 on Build GRCh38
rs1017207606 Chr.17: 40555582-40555582 on Build GRCh38
rs199621619 Chr.17: 40555525-40555525 on Build GRCh38
rs897792209 Chr.17: 40555576-40555576 on Build GRCh38
rs761681023 Chr.17: 40555565-40555565 on Build GRCh38

View Full Product Details