Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 207733886-207734113 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNYL1
Entrez Gene ID
151195
Gene Name
cyclin Y like 1
Gene Aliases
-
UniGene
Hs.471234
Gene Chromosome Location
Chr.2: 207711540-207756174 on Build GRCh38
Transcripts
NM_001142300.1 NM_152523.2 XM_005246343.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4953286 c.193C>T p.Q65* Chr.2: 207734019-207734019 on Build GRCh38
6581456 c.136C>T p.L46F Chr.2: 207733962-207733962 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs752287007 Chr.2: 207734070-207734070 on Build GRCh38
rs773104179 Chr.2: 207733959-207733959 on Build GRCh38
rs35335346 Chr.2: 207733967-207733968 on Build GRCh38
rs376783839 Chr.2: 207733966-207733966 on Build GRCh38
rs751997670 Chr.2: 207734083-207734083 on Build GRCh38
rs991866251 Chr.2: 207734045-207734045 on Build GRCh38
rs374291612 Chr.2: 207734081-207734081 on Build GRCh38
rs371230375 Chr.2: 207733992-207733992 on Build GRCh38
rs201263386 Chr.2: 207733939-207733939 on Build GRCh38
rs759057580 Chr.2: 207734040-207734040 on Build GRCh38
rs769200164 Chr.2: 207734084-207734084 on Build GRCh38
rs771747848 Chr.2: 207733933-207733933 on Build GRCh38
rs781771006 Chr.2: 207734088-207734088 on Build GRCh38
rs960882544 Chr.2: 207734002-207734002 on Build GRCh38
rs766332715 Chr.2: 207733997-207733997 on Build GRCh38
rs758129104 Chr.2: 207734072-207734072 on Build GRCh38
rs749087320 Chr.2: 207733944-207733944 on Build GRCh38
rs991258302 Chr.2: 207733998-207733998 on Build GRCh38
rs930762939 Chr.2: 207734086-207734086 on Build GRCh38
rs774164145 Chr.2: 207733965-207733965 on Build GRCh38
rs764803405 Chr.2: 207734069-207734069 on Build GRCh38

View Full Product Details