Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 104938829-104939066 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AHNAK2
Entrez Gene ID
113146
Gene Name
AHNAK nucleoprotein 2
Gene Aliases
C14orf78
UniGene
Hs.441783
Gene Chromosome Location
Chr.14: 104937253-104978357 on Build GRCh38
Transcripts
NM_138420.2 XM_005267299.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5971063 c.1301G>C p.R434T Chr.14: 104939060-104939060 on Build GRCh38
697859 c.16518C>A p.I5506I Chr.14: 104938933-104938933 on Build GRCh38
6838715 c.1384G>C p.V462L Chr.14: 104938977-104938977 on Build GRCh38
5074635 c.16544C>T p.S5515L Chr.14: 104938907-104938907 on Build GRCh38
6841695 c.1499C>T p.T500M Chr.14: 104938862-104938862 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760312397 Chr.14: 104938856-104938856 on Build GRCh38
rs752660844 Chr.14: 104938873-104938873 on Build GRCh38
rs377580543 Chr.14: 104938947-104938947 on Build GRCh38
rs771202267 Chr.14: 104938945-104938945 on Build GRCh38
rs749506720 Chr.14: 104938920-104938920 on Build GRCh38
rs984705249 Chr.14: 104939028-104939028 on Build GRCh38
rs763978102 Chr.14: 104938903-104938903 on Build GRCh38
rs780257891 Chr.14: 104938915-104938915 on Build GRCh38
rs1052749191 Chr.14: 104938879-104938879 on Build GRCh38
rs755193540 Chr.14: 104939044-104939044 on Build GRCh38
rs754065940 Chr.14: 104939037-104939037 on Build GRCh38
rs200811905 Chr.14: 104938946-104938946 on Build GRCh38
rs563792120 Chr.14: 104938864-104938864 on Build GRCh38
rs762970824 Chr.14: 104938972-104938972 on Build GRCh38
rs974107476 Chr.14: 104939030-104939030 on Build GRCh38
rs529291911 Chr.14: 104938931-104938931 on Build GRCh38
rs1052215012 Chr.14: 104938884-104938884 on Build GRCh38
rs767242374 Chr.14: 104939024-104939024 on Build GRCh38
rs868698057 Chr.14: 104939020-104939020 on Build GRCh38
rs200712744 Chr.14: 104938906-104938906 on Build GRCh38
rs140424316 Chr.14: 104938862-104938862 on Build GRCh38
rs748618240 Chr.14: 104938929-104938929 on Build GRCh38
rs566012025 Chr.14: 104938943-104938943 on Build GRCh38
rs1002952394 Chr.14: 104938866-104938866 on Build GRCh38
rs781533202 Chr.14: 104939031-104939032 on Build GRCh38
rs372998948 Chr.14: 104938928-104938928 on Build GRCh38
rs755393516 Chr.14: 104938928-104938929 on Build GRCh38
rs758449903 Chr.14: 104938902-104938902 on Build GRCh38
rs764070003 Chr.14: 104938985-104938985 on Build GRCh38
rs976496428 Chr.14: 104938975-104938975 on Build GRCh38
rs201885720 Chr.14: 104938950-104938950 on Build GRCh38
rs773551057 Chr.14: 104938933-104938933 on Build GRCh38
rs751552545 Chr.14: 104938992-104938992 on Build GRCh38
rs892118601 Chr.14: 104938895-104938895 on Build GRCh38
rs1022905008 Chr.14: 104938927-104938927 on Build GRCh38
rs749248609 Chr.14: 104938964-104938964 on Build GRCh38
rs757019628 Chr.14: 104938907-104938907 on Build GRCh38
rs766113093 Chr.14: 104938860-104938860 on Build GRCh38
rs776161477 Chr.14: 104938861-104938861 on Build GRCh38
rs761631310 Chr.14: 104939007-104939007 on Build GRCh38
rs375860743 Chr.14: 104939027-104939027 on Build GRCh38

View Full Product Details