Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 120136577-120136802 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCN1
Entrez Gene ID
10985
Gene Name
GCN1, eIF2 alpha kinase activator homolog
Gene Aliases
GCN1L, GCN1L1, PRIC295
UniGene
Hs.298716
Gene Chromosome Location
Chr.12: 120127210-120194710 on Build GRCh38
Transcripts
NM_006836.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
272718 c.6859T>C p.L2287L Chr.12: 120136651-120136651 on Build GRCh38
5531637 c.6905C>T p.S2302F Chr.12: 120136605-120136605 on Build GRCh38
467885 c.6848C>T p.A2283V Chr.12: 120136662-120136662 on Build GRCh38
219405 c.6857C>T p.A2286V Chr.12: 120136653-120136653 on Build GRCh38
3456987 c.6933C>T p.I2311I Chr.12: 120136577-120136577 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771890293 Chr.12: 120136626-120136626 on Build GRCh38
rs777529738 Chr.12: 120136603-120136603 on Build GRCh38
rs760382222 Chr.12: 120136770-120136770 on Build GRCh38
rs777920038 Chr.12: 120136729-120136729 on Build GRCh38
rs775044229 Chr.12: 120136628-120136628 on Build GRCh38
rs933405209 Chr.12: 120136696-120136696 on Build GRCh38
rs200128356 Chr.12: 120136636-120136636 on Build GRCh38
rs544434380 Chr.12: 120136646-120136646 on Build GRCh38
rs755166395 Chr.12: 120136677-120136677 on Build GRCh38
rs375505009 Chr.12: 120136779-120136779 on Build GRCh38
rs200758132 Chr.12: 120136737-120136737 on Build GRCh38
rs771058175 Chr.12: 120136739-120136739 on Build GRCh38
rs745707357 Chr.12: 120136741-120136741 on Build GRCh38
rs866096088 Chr.12: 120136624-120136624 on Build GRCh38
rs370364196 Chr.12: 120136605-120136605 on Build GRCh38
rs199987142 Chr.12: 120136604-120136604 on Build GRCh38
rs200087555 Chr.12: 120136740-120136740 on Build GRCh38
rs1050416777 Chr.12: 120136698-120136698 on Build GRCh38
rs756078676 Chr.12: 120136717-120136717 on Build GRCh38
rs774344398 Chr.12: 120136618-120136618 on Build GRCh38
rs759256305 Chr.12: 120136623-120136623 on Build GRCh38
rs760056183 Chr.12: 120136633-120136633 on Build GRCh38
rs763450350 Chr.12: 120136781-120136781 on Build GRCh38
rs772015009 Chr.12: 120136752-120136752 on Build GRCh38
rs201082049 Chr.12: 120136635-120136635 on Build GRCh38
rs762365859 Chr.12: 120136751-120136763 on Build GRCh38
rs867977828 Chr.12: 120136637-120136637 on Build GRCh38
rs377583315 Chr.12: 120136767-120136767 on Build GRCh38
rs781218382 Chr.12: 120136694-120136694 on Build GRCh38
rs769935640 Chr.12: 120136600-120136600 on Build GRCh38
rs367654754 Chr.12: 120136703-120136703 on Build GRCh38
rs766719757 Chr.12: 120136649-120136649 on Build GRCh38
rs767995376 Chr.12: 120136772-120136772 on Build GRCh38

View Full Product Details