Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 6622537-6622810 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCHS1
Entrez Gene ID
8642
Gene Name
dachsous cadherin-related 1
Gene Aliases
CDH19, CDH25, CDHR6, FIB1, MVP2, PCDH16, VMLDS1
UniGene
Hs.199850
Gene Chromosome Location
Chr.11: 6621324-6655849 on Build GRCh38
Transcripts
NM_003737.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7297407 c.9123T>C p.D3041D Chr.11: 6622553-6622553 on Build GRCh38
5475561 c.8972G>A p.R2991Q Chr.11: 6622704-6622704 on Build GRCh38
3752669 c.8931G>A p.Q2977Q Chr.11: 6622745-6622745 on Build GRCh38
5721660 c.9061C>T p.R3021C Chr.11: 6622615-6622615 on Build GRCh38
5492751 c.8922A>G p.P2974P Chr.11: 6622754-6622754 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369667426 Chr.11: 6622756-6622756 on Build GRCh38
rs749130141 Chr.11: 6622697-6622697 on Build GRCh38
rs780210754 Chr.11: 6622563-6622571 on Build GRCh38
rs1039088358 Chr.11: 6622630-6622630 on Build GRCh38
rs780578129 Chr.11: 6622675-6622675 on Build GRCh38
rs749365833 Chr.11: 6622747-6622748 on Build GRCh38
rs771972771 Chr.11: 6622786-6622786 on Build GRCh38
rs893819999 Chr.11: 6622632-6622632 on Build GRCh38
rs763425383 Chr.11: 6622739-6622739 on Build GRCh38
rs912004965 Chr.11: 6622561-6622561 on Build GRCh38
rs765898503 Chr.11: 6622658-6622658 on Build GRCh38
rs548201105 Chr.11: 6622784-6622784 on Build GRCh38
rs947274722 Chr.11: 6622637-6622637 on Build GRCh38
rs536194145 Chr.11: 6622782-6622782 on Build GRCh38
rs777372556 Chr.11: 6622696-6622696 on Build GRCh38
rs7122587 Chr.11: 6622745-6622745 on Build GRCh38
rs760992622 Chr.11: 6622570-6622570 on Build GRCh38
rs550669430 Chr.11: 6622668-6622668 on Build GRCh38
rs753521014 Chr.11: 6622780-6622780 on Build GRCh38
rs758870230 Chr.11: 6622672-6622672 on Build GRCh38
rs201519860 Chr.11: 6622779-6622779 on Build GRCh38
rs764454482 Chr.11: 6622724-6622724 on Build GRCh38
rs867227843 Chr.11: 6622578-6622578 on Build GRCh38
rs780013390 Chr.11: 6622788-6622788 on Build GRCh38
rs988875015 Chr.11: 6622583-6622583 on Build GRCh38
rs533377974 Chr.11: 6622727-6622727 on Build GRCh38
rs760836404 Chr.11: 6622721-6622721 on Build GRCh38
rs775492992 Chr.11: 6622717-6622717 on Build GRCh38
rs757668517 Chr.11: 6622652-6622652 on Build GRCh38
rs745798498 Chr.11: 6622705-6622705 on Build GRCh38
rs752243064 Chr.11: 6622758-6622758 on Build GRCh38
rs755830203 Chr.11: 6622683-6622683 on Build GRCh38
rs200152314 Chr.11: 6622704-6622704 on Build GRCh38
rs200940363 Chr.11: 6622785-6622785 on Build GRCh38
rs754324316 Chr.11: 6622650-6622650 on Build GRCh38
rs867539218 Chr.11: 6622633-6622633 on Build GRCh38
rs750737940 Chr.11: 6622737-6622737 on Build GRCh38
rs377192523 Chr.11: 6622716-6622716 on Build GRCh38
rs764401147 Chr.11: 6622599-6622599 on Build GRCh38
rs374516352 Chr.11: 6622682-6622682 on Build GRCh38
rs943349167 Chr.11: 6622614-6622614 on Build GRCh38
rs762082110 Chr.11: 6622735-6622735 on Build GRCh38
rs781632057 Chr.11: 6622733-6622733 on Build GRCh38
rs140276142 Chr.11: 6622775-6622775 on Build GRCh38

View Full Product Details