Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 158256879-158257112 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD1A
Entrez Gene ID
909
Gene Name
CD1a molecule
Gene Aliases
CD1, FCB6, HTA1, R4, T6
UniGene
Hs.1309
Gene Chromosome Location
Chr.1: 158248329-158258269 on Build GRCh38
Transcripts
NM_001320652.1 NM_001763.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
391299 c.811_812delGG p.E272fs*3 Chr.1: 158256992-158256993 on Build GRCh38
3863118 c.762C>T p.P254P Chr.1: 158256943-158256943 on Build GRCh38
389810 c.836G>A p.R279Q Chr.1: 158257017-158257017 on Build GRCh38
296748 c.790G>A p.A264T Chr.1: 158256971-158256971 on Build GRCh38
1601021 c.811G>T p.G271W Chr.1: 158256992-158256992 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751121536 Chr.1: 158256941-158256941 on Build GRCh38
rs761269454 Chr.1: 158257035-158257035 on Build GRCh38
rs41310138 Chr.1: 158257024-158257024 on Build GRCh38
rs570589065 Chr.1: 158257065-158257065 on Build GRCh38
rs764472035 Chr.1: 158257038-158257038 on Build GRCh38
rs778421503 Chr.1: 158256910-158256910 on Build GRCh38
rs745499344 Chr.1: 158256988-158256988 on Build GRCh38
rs746460800 Chr.1: 158257056-158257056 on Build GRCh38
rs1045671736 Chr.1: 158257055-158257055 on Build GRCh38
rs757980794 Chr.1: 158257051-158257051 on Build GRCh38
rs920808231 Chr.1: 158257077-158257077 on Build GRCh38
rs759735425 Chr.1: 158256957-158256957 on Build GRCh38
rs759825385 Chr.1: 158257020-158257020 on Build GRCh38
rs894156813 Chr.1: 158257045-158257045 on Build GRCh38
rs369993390 Chr.1: 158256929-158256929 on Build GRCh38
rs775376808 Chr.1: 158257028-158257028 on Build GRCh38
rs377049912 Chr.1: 158256899-158256899 on Build GRCh38
rs750143676 Chr.1: 158257046-158257046 on Build GRCh38
rs894951610 Chr.1: 158256977-158256977 on Build GRCh38
rs141060322 Chr.1: 158256900-158256900 on Build GRCh38
rs774767602 Chr.1: 158257019-158257019 on Build GRCh38
rs140895583 Chr.1: 158257058-158257058 on Build GRCh38
rs1004083885 Chr.1: 158257059-158257059 on Build GRCh38
rs369997586 Chr.1: 158257044-158257044 on Build GRCh38
rs373354671 Chr.1: 158256951-158256951 on Build GRCh38
rs781040103 Chr.1: 158257062-158257062 on Build GRCh38
rs772374326 Chr.1: 158257090-158257090 on Build GRCh38
rs149659983 Chr.1: 158256926-158256926 on Build GRCh38
rs778580983 Chr.1: 158256982-158256982 on Build GRCh38
rs149395316 Chr.1: 158257064-158257064 on Build GRCh38
rs143177914 Chr.1: 158256989-158256989 on Build GRCh38
rs746978676 Chr.1: 158257005-158257005 on Build GRCh38
rs766668283 Chr.1: 158256938-158256938 on Build GRCh38
rs914038336 Chr.1: 158257003-158257003 on Build GRCh38
rs372371602 Chr.1: 158257082-158257082 on Build GRCh38
rs140268166 Chr.1: 158256927-158256927 on Build GRCh38
rs141426527 Chr.1: 158257016-158257016 on Build GRCh38
rs768426578 Chr.1: 158257009-158257009 on Build GRCh38
rs756341916 Chr.1: 158256932-158256932 on Build GRCh38
rs780179448 Chr.1: 158256953-158256953 on Build GRCh38
rs920848395 Chr.1: 158256949-158256949 on Build GRCh38
rs189152658 Chr.1: 158257042-158257042 on Build GRCh38
rs779636773 Chr.1: 158257052-158257052 on Build GRCh38
rs1028337498 Chr.1: 158256985-158256985 on Build GRCh38
rs756938435 Chr.1: 158256974-158256974 on Build GRCh38
rs746371418 Chr.1: 158257043-158257043 on Build GRCh38
rs569199745 Chr.1: 158256908-158256908 on Build GRCh38
rs140989108 Chr.1: 158256969-158256969 on Build GRCh38
rs754343758 Chr.1: 158257040-158257040 on Build GRCh38
rs777992461 Chr.1: 158256905-158256905 on Build GRCh38
rs141143133 Chr.1: 158256968-158256968 on Build GRCh38
rs778216345 Chr.1: 158256971-158256971 on Build GRCh38
rs769775413 Chr.1: 158257073-158257073 on Build GRCh38
rs898542231 Chr.1: 158257076-158257076 on Build GRCh38
rs376991873 Chr.1: 158256970-158256970 on Build GRCh38
rs746099693 Chr.1: 158257085-158257085 on Build GRCh38
rs745346466 Chr.1: 158256915-158256915 on Build GRCh38
rs781773699 Chr.1: 158256897-158256897 on Build GRCh38
rs1012509994 Chr.1: 158257006-158257006 on Build GRCh38
rs769686492 Chr.1: 158257017-158257017 on Build GRCh38

View Full Product Details