Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64118172-64118397 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLRT1
Entrez Gene ID
23769
Gene Name
fibronectin leucine rich transmembrane protein 1
Gene Aliases
SPG68
UniGene
Hs.584876
Gene Chromosome Location
Chr.11: 64035970-64119183 on Build GRCh38
Transcripts
NM_013280.4 XM_005273861.1 XM_011544883.1 NM_014067.3 XM_005273939.4 XM_006718521.3 XM_011544970.2

Gene Symbol
MACROD1
Entrez Gene ID
28992
Gene Name
MACRO domain containing 1
Gene Aliases
LRP16
UniGene
Hs.602898
Gene Chromosome Location
Chr.11: 63998554-64166652 on Build GRCh38
Transcripts
NM_013280.4 XM_005273861.1 XM_011544883.1 NM_014067.3 XM_005273939.4 XM_006718521.3 XM_011544970.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3451371 c.1987G>A p.D663N Chr.11: 64118254-64118254 on Build GRCh38
6665160 c.1994G>A p.G665D Chr.11: 64118261-64118261 on Build GRCh38
5360733 c.1984C>A p.R662R Chr.11: 64118251-64118251 on Build GRCh38
1604946 c.1926C>T p.N642N Chr.11: 64118193-64118193 on Build GRCh38
4889318 c.2016C>A p.S672S Chr.11: 64118283-64118283 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1047952824 Chr.11: 64118301-64118301 on Build GRCh38
rs971779587 Chr.11: 64118343-64118343 on Build GRCh38
rs767651641 Chr.11: 64118316-64118316 on Build GRCh38
rs763052835 Chr.11: 64118299-64118299 on Build GRCh38
rs757351313 Chr.11: 64118243-64118243 on Build GRCh38
rs771473912 Chr.11: 64118207-64118207 on Build GRCh38
rs777319814 Chr.11: 64118268-64118268 on Build GRCh38
rs369302404 Chr.11: 64118306-64118306 on Build GRCh38
rs746475638 Chr.11: 64118269-64118269 on Build GRCh38
rs372963657 Chr.11: 64118251-64118251 on Build GRCh38
rs181750370 Chr.11: 64118315-64118315 on Build GRCh38
rs750921137 Chr.11: 64118318-64118318 on Build GRCh38
rs201346437 Chr.11: 64118298-64118298 on Build GRCh38
rs79628679 Chr.11: 64118225-64118225 on Build GRCh38
rs750934842 Chr.11: 64118252-64118252 on Build GRCh38
rs746183688 Chr.11: 64118194-64118194 on Build GRCh38
rs550871684 Chr.11: 64118273-64118273 on Build GRCh38
rs772829544 Chr.11: 64118213-64118213 on Build GRCh38
rs771799567 Chr.11: 64118266-64118266 on Build GRCh38
rs948207838 Chr.11: 64118218-64118218 on Build GRCh38
rs566188754 Chr.11: 64118203-64118203 on Build GRCh38
rs752857407 Chr.11: 64118239-64118239 on Build GRCh38
rs764056550 Chr.11: 64118241-64118241 on Build GRCh38
rs770157471 Chr.11: 64118198-64118198 on Build GRCh38
rs973817480 Chr.11: 64118288-64118288 on Build GRCh38
rs888654575 Chr.11: 64118195-64118195 on Build GRCh38
rs756470729 Chr.11: 64118255-64118255 on Build GRCh38
rs150772900 Chr.11: 64118242-64118242 on Build GRCh38
rs200253771 Chr.11: 64118324-64118324 on Build GRCh38
rs747737126 Chr.11: 64118260-64118260 on Build GRCh38
rs745867021 Chr.11: 64118290-64118290 on Build GRCh38
rs528544288 Chr.11: 64118219-64118219 on Build GRCh38
rs372484626 Chr.11: 64118193-64118193 on Build GRCh38
rs537238642 Chr.11: 64118240-64118240 on Build GRCh38
rs918956445 Chr.11: 64118365-64118365 on Build GRCh38
rs765128867 Chr.11: 64118233-64118233 on Build GRCh38
rs930328080 Chr.11: 64118366-64118366 on Build GRCh38
rs535222834 Chr.11: 64118339-64118339 on Build GRCh38
rs372560428 Chr.11: 64118220-64118220 on Build GRCh38
rs677447 Chr.11: 64118232-64118232 on Build GRCh38
rs688241 Chr.11: 64118297-64118297 on Build GRCh38
rs766624364 Chr.11: 64118325-64118325 on Build GRCh38
rs780458547 Chr.11: 64118257-64118257 on Build GRCh38
rs779412000 Chr.11: 64118289-64118290 on Build GRCh38
rs754188520 Chr.11: 64118338-64118338 on Build GRCh38
rs776639848 Chr.11: 64118278-64118278 on Build GRCh38

View Full Product Details