Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 34867433-34867704 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLGAP3
Entrez Gene ID
58512
Gene Name
DLG associated protein 3
Gene Aliases
DAP3, SAPAP3
UniGene
Hs.733945
Gene Chromosome Location
Chr.1: 34862120-34929585 on Build GRCh38
Transcripts
NM_001080418.2 XM_011541879.2 XM_011541880.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1342081 c.2488C>A p.L830M Chr.1: 34867625-34867625 on Build GRCh38
5380783 c.2566C>T p.Q856* Chr.1: 34867547-34867547 on Build GRCh38
1342080 c.2553C>A p.F851L Chr.1: 34867560-34867560 on Build GRCh38
6644976 c.2560C>T p.L854L Chr.1: 34867553-34867553 on Build GRCh38
4397045 c.2545C>T p.Q849* Chr.1: 34867568-34867568 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747513748 Chr.1: 34867674-34867674 on Build GRCh38
rs750746536 Chr.1: 34867663-34867663 on Build GRCh38
rs181842419 Chr.1: 34867509-34867509 on Build GRCh38
rs899423995 Chr.1: 34867640-34867640 on Build GRCh38
rs941464858 Chr.1: 34867495-34867495 on Build GRCh38
rs773111340 Chr.1: 34867610-34867610 on Build GRCh38
rs776457680 Chr.1: 34867571-34867571 on Build GRCh38
rs777127004 Chr.1: 34867586-34867586 on Build GRCh38
rs769763442 Chr.1: 34867598-34867598 on Build GRCh38
rs12727066 Chr.1: 34867525-34867525 on Build GRCh38
rs372791422 Chr.1: 34867646-34867646 on Build GRCh38
rs746484707 Chr.1: 34867556-34867556 on Build GRCh38
rs754081612 Chr.1: 34867655-34867655 on Build GRCh38
rs374534747 Chr.1: 34867493-34867493 on Build GRCh38
rs540159412 Chr.1: 34867555-34867555 on Build GRCh38
rs371640894 Chr.1: 34867667-34867667 on Build GRCh38
rs748658137 Chr.1: 34867612-34867612 on Build GRCh38
rs758998680 Chr.1: 34867664-34867664 on Build GRCh38
rs763014828 Chr.1: 34867617-34867617 on Build GRCh38
rs761511876 Chr.1: 34867578-34867578 on Build GRCh38
rs79950652 Chr.1: 34867657-34867657 on Build GRCh38
rs57077677 Chr.1: 34867654-34867654 on Build GRCh38
rs777983766 Chr.1: 34867488-34867488 on Build GRCh38
rs924567010 Chr.1: 34867466-34867466 on Build GRCh38
rs766214978 Chr.1: 34867650-34867650 on Build GRCh38
rs371385053 Chr.1: 34867560-34867560 on Build GRCh38
rs933408381 Chr.1: 34867480-34867480 on Build GRCh38
rs551307690 Chr.1: 34867528-34867528 on Build GRCh38
rs751455541 Chr.1: 34867651-34867651 on Build GRCh38
rs753991553 Chr.1: 34867491-34867491 on Build GRCh38
rs779286632 Chr.1: 34867661-34867661 on Build GRCh38

View Full Product Details