Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 108133982-108134233 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG4A
Entrez Gene ID
115201
Gene Name
autophagy related 4A cysteine peptidase
Gene Aliases
APG4A, AUTL2
UniGene
Hs.8763
Gene Chromosome Location
Chr.X: 108088900-108154671 on Build GRCh38
Transcripts
NM_001321287.1 NM_001321288.1 NM_001321289.1 NM_001321290.1 NM_052936.4 NM_178270.3 NM_178271.2 NR_135608.1 XM_011530841.2 XM_011530842.1

SNP Detail

SNP ID Chromosome Location
rs771847676 Chr.X: 108134082-108134082 on Build GRCh38
rs774766454 Chr.X: 108134166-108134166 on Build GRCh38
rs766341518 Chr.X: 108134176-108134176 on Build GRCh38
rs748457649 Chr.X: 108134158-108134158 on Build GRCh38
rs373701323 Chr.X: 108134205-108134205 on Build GRCh38
rs757931045 Chr.X: 108134038-108134039 on Build GRCh38
rs776735899 Chr.X: 108134181-108134181 on Build GRCh38
rs369180626 Chr.X: 108134175-108134175 on Build GRCh38
rs774160022 Chr.X: 108134102-108134102 on Build GRCh38
rs768305363 Chr.X: 108134044-108134044 on Build GRCh38
rs752979182 Chr.X: 108134204-108134204 on Build GRCh38
rs374845036 Chr.X: 108134039-108134039 on Build GRCh38
rs1041310812 Chr.X: 108134022-108134022 on Build GRCh38
rs1028787561 Chr.X: 108134149-108134149 on Build GRCh38
rs767347050 Chr.X: 108134206-108134206 on Build GRCh38
rs759561476 Chr.X: 108134191-108134191 on Build GRCh38
rs772632658 Chr.X: 108134163-108134163 on Build GRCh38
rs765446781 Chr.X: 108134201-108134201 on Build GRCh38
rs762465351 Chr.X: 108134039-108134039 on Build GRCh38
rs779653587 Chr.X: 108134027-108134027 on Build GRCh38

View Full Product Details