Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55691893-55692122 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPN1
Entrez Gene ID
29924
Gene Name
epsin 1
Gene Aliases
-
UniGene
Hs.279953
Gene Chromosome Location
Chr.19: 55675195-55695767 on Build GRCh38
Transcripts
NM_001130071.1 NM_001130072.1 NM_013333.3 XM_005258829.2 XM_005258830.2 XM_011526880.1 XM_011526881.1 XM_017026722.1 XM_017026723.1 XM_017026724.1 XM_017026725.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6384948 c.920C>A p.P307H Chr.19: 55691911-55691911 on Build GRCh38
6384947 c.1178C>A p.P393H Chr.19: 55691911-55691911 on Build GRCh38
7275462 c.930T>C p.G310G Chr.19: 55691921-55691921 on Build GRCh38
3539500 c.904C>T p.P302S Chr.19: 55691895-55691895 on Build GRCh38
5578489 c.958C>T p.P320S Chr.19: 55691949-55691949 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs534258974 Chr.19: 55691922-55691922 on Build GRCh38
rs755550490 Chr.19: 55692002-55692002 on Build GRCh38
rs185413887 Chr.19: 55692064-55692064 on Build GRCh38
rs767077051 Chr.19: 55692087-55692087 on Build GRCh38
rs1032392272 Chr.19: 55692092-55692092 on Build GRCh38
rs766998303 Chr.19: 55691967-55691967 on Build GRCh38
rs750465690 Chr.19: 55691964-55691964 on Build GRCh38
rs779995954 Chr.19: 55692004-55692004 on Build GRCh38
rs745379863 Chr.19: 55691986-55691986 on Build GRCh38
rs751267822 Chr.19: 55691975-55691975 on Build GRCh38
rs765676193 Chr.19: 55691970-55691970 on Build GRCh38
rs768488143 Chr.19: 55692014-55692014 on Build GRCh38
rs755640442 Chr.19: 55692093-55692093 on Build GRCh38
rs568366532 Chr.19: 55691916-55691916 on Build GRCh38
rs769608556 Chr.19: 55691927-55691927 on Build GRCh38
rs761301947 Chr.19: 55691966-55691966 on Build GRCh38
rs935171451 Chr.19: 55692007-55692007 on Build GRCh38
rs756858015 Chr.19: 55691979-55691979 on Build GRCh38
rs193174793 Chr.19: 55691926-55691926 on Build GRCh38
rs758735504 Chr.19: 55692072-55692072 on Build GRCh38
rs771574556 Chr.19: 55692065-55692065 on Build GRCh38
rs375632073 Chr.19: 55692054-55692054 on Build GRCh38
rs1017922901 Chr.19: 55692070-55692070 on Build GRCh38
rs768946948 Chr.19: 55691926-55691928 on Build GRCh38
rs544835123 Chr.19: 55691983-55691983 on Build GRCh38
rs749171270 Chr.19: 55692008-55692008 on Build GRCh38
rs201565739 Chr.19: 55691935-55691935 on Build GRCh38
rs761917774 Chr.19: 55691936-55691936 on Build GRCh38
rs558415640 Chr.19: 55692053-55692053 on Build GRCh38
rs576899143 Chr.19: 55692091-55692091 on Build GRCh38
rs374441106 Chr.19: 55692066-55692066 on Build GRCh38
rs754269275 Chr.19: 55691910-55691910 on Build GRCh38
rs775345018 Chr.19: 55691929-55691929 on Build GRCh38
rs567418507 Chr.19: 55691965-55691965 on Build GRCh38
rs755327224 Chr.19: 55691913-55691913 on Build GRCh38
rs755417216 Chr.19: 55691968-55691968 on Build GRCh38
rs937790888 Chr.19: 55692027-55692027 on Build GRCh38
rs1041064576 Chr.19: 55692071-55692071 on Build GRCh38
rs768398238 Chr.19: 55691933-55691933 on Build GRCh38
rs866892047 Chr.19: 55692057-55692057 on Build GRCh38
rs772170929 Chr.19: 55692031-55692031 on Build GRCh38
rs760701138 Chr.19: 55692039-55692039 on Build GRCh38
rs766520788 Chr.19: 55692049-55692049 on Build GRCh38
rs767673424 Chr.19: 55691946-55691946 on Build GRCh38
rs1032528084 Chr.19: 55691915-55691915 on Build GRCh38
rs369334871 Chr.19: 55691920-55691920 on Build GRCh38
rs780539299 Chr.19: 55691923-55691923 on Build GRCh38
rs748470203 Chr.19: 55692018-55692018 on Build GRCh38
rs951128642 Chr.19: 55692101-55692101 on Build GRCh38
rs762737827 Chr.19: 55691932-55691932 on Build GRCh38

View Full Product Details