Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 47977300-47977573 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BSPH1
Entrez Gene ID
100131137
Gene Name
binder of sperm protein homolog 1
Gene Aliases
BSP1, ELSPBP2
UniGene
Hs.651072
Gene Chromosome Location
Chr.19: 47967258-47992170 on Build GRCh38
Transcripts
NM_001128326.2 XM_017026118.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4891704 c.201G>T p.K67N Chr.19: 47977428-47977428 on Build GRCh38
6847125 c.128G>C p.G43A Chr.19: 47977501-47977501 on Build GRCh38
5707102 c.169_171delTAT p.Y57delY Chr.19: 47977458-47977460 on Build GRCh38
5936431 c.226G>A p.E76K Chr.19: 47977403-47977403 on Build GRCh38
5891042 c.161G>A p.G54E Chr.19: 47977468-47977468 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs926739822 Chr.19: 47977322-47977322 on Build GRCh38
rs903996391 Chr.19: 47977411-47977411 on Build GRCh38
rs776330351 Chr.19: 47977370-47977370 on Build GRCh38
rs746437945 Chr.19: 47977493-47977493 on Build GRCh38
rs535097244 Chr.19: 47977465-47977465 on Build GRCh38
rs373760536 Chr.19: 47977379-47977379 on Build GRCh38
rs966606956 Chr.19: 47977403-47977403 on Build GRCh38
rs552626585 Chr.19: 47977450-47977450 on Build GRCh38
rs1039640919 Chr.19: 47977423-47977423 on Build GRCh38
rs772685776 Chr.19: 47977529-47977529 on Build GRCh38
rs1042621385 Chr.19: 47977395-47977395 on Build GRCh38
rs958251000 Chr.19: 47977453-47977453 on Build GRCh38
rs899565126 Chr.19: 47977520-47977520 on Build GRCh38
rs908005939 Chr.19: 47977532-47977532 on Build GRCh38
rs190252446 Chr.19: 47977320-47977320 on Build GRCh38
rs938121498 Chr.19: 47977333-47977333 on Build GRCh38
rs560757821 Chr.19: 47977492-47977493 on Build GRCh38
rs185872432 Chr.19: 47977459-47977459 on Build GRCh38
rs997041688 Chr.19: 47977525-47977525 on Build GRCh38
rs990824013 Chr.19: 47977455-47977455 on Build GRCh38
rs949448775 Chr.19: 47977504-47977504 on Build GRCh38
rs764826403 Chr.19: 47977538-47977538 on Build GRCh38
rs773357990 Chr.19: 47977344-47977344 on Build GRCh38
rs375584891 Chr.19: 47977392-47977392 on Build GRCh38
rs150415301 Chr.19: 47977419-47977419 on Build GRCh38
rs1019402441 Chr.19: 47977380-47977380 on Build GRCh38
rs555389557 Chr.19: 47977514-47977514 on Build GRCh38
rs180992446 Chr.19: 47977346-47977346 on Build GRCh38
rs764447546 Chr.19: 47977505-47977505 on Build GRCh38
rs774572875 Chr.19: 47977352-47977352 on Build GRCh38
rs945598730 Chr.19: 47977363-47977363 on Build GRCh38
rs770995161 Chr.19: 47977356-47977356 on Build GRCh38

View Full Product Details