Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 14819869-14820140 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDNF
Entrez Gene ID
441549
Gene Name
cerebral dopamine neurotrophic factor
Gene Aliases
ARMETL1
UniGene
Hs.559067
Gene Chromosome Location
Chr.10: 14819245-14838073 on Build GRCh38
Transcripts
NM_001029954.2 XM_011519488.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7293804 c.519G>A p.E173E Chr.10: 14820025-14820025 on Build GRCh38
5890475 c.514C>T p.Q172* Chr.10: 14820030-14820030 on Build GRCh38
5902945 c.526C>T p.P176S Chr.10: 14820018-14820018 on Build GRCh38
4612258 c.493_494insA p.T165fs*2 Chr.10: 14820050-14820051 on Build GRCh38
1162904 c.551A>G p.K184R Chr.10: 14819993-14819993 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759972267 Chr.10: 14819950-14819950 on Build GRCh38
rs367910630 Chr.10: 14820082-14820082 on Build GRCh38
rs140598117 Chr.10: 14819961-14819961 on Build GRCh38
rs761847471 Chr.10: 14820069-14820069 on Build GRCh38
rs748926273 Chr.10: 14820014-14820014 on Build GRCh38
rs945451378 Chr.10: 14820101-14820101 on Build GRCh38
rs762016430 Chr.10: 14820085-14820086 on Build GRCh38
rs771777537 Chr.10: 14820034-14820034 on Build GRCh38
rs764168746 Chr.10: 14819985-14819985 on Build GRCh38
rs61738953 Chr.10: 14820083-14820083 on Build GRCh38
rs372311274 Chr.10: 14819945-14819945 on Build GRCh38
rs747844191 Chr.10: 14820106-14820106 on Build GRCh38
rs61746971 Chr.10: 14820043-14820043 on Build GRCh38
rs367858997 Chr.10: 14820003-14820003 on Build GRCh38
rs775722131 Chr.10: 14820046-14820046 on Build GRCh38
rs745585947 Chr.10: 14819989-14819998 on Build GRCh38
rs757383518 Chr.10: 14819998-14819998 on Build GRCh38
rs112164059 Chr.10: 14819978-14819978 on Build GRCh38
rs764286370 Chr.10: 14820061-14820061 on Build GRCh38
rs920235044 Chr.10: 14819973-14819973 on Build GRCh38
rs776483572 Chr.10: 14819940-14819943 on Build GRCh38
rs763106051 Chr.10: 14819980-14819980 on Build GRCh38
rs778511317 Chr.10: 14820021-14820021 on Build GRCh38
rs754864330 Chr.10: 14820084-14820084 on Build GRCh38
rs747650414 Chr.10: 14820032-14820032 on Build GRCh38
rs767629550 Chr.10: 14820078-14820078 on Build GRCh38
rs762047925 Chr.10: 14820004-14820004 on Build GRCh38
rs770147902 Chr.10: 14820044-14820044 on Build GRCh38
rs769017633 Chr.10: 14820043-14820044 on Build GRCh38
rs774667158 Chr.10: 14820050-14820050 on Build GRCh38
rs751651711 Chr.10: 14819986-14819986 on Build GRCh38
rs754595681 Chr.10: 14820017-14820017 on Build GRCh38
rs763176247 Chr.10: 14820059-14820059 on Build GRCh38
rs761194597 Chr.10: 14819931-14819931 on Build GRCh38
rs771575286 Chr.10: 14819934-14819934 on Build GRCh38
rs1018742163 Chr.10: 14819928-14819928 on Build GRCh38
rs956718529 Chr.10: 14820105-14820105 on Build GRCh38
rs777332825 Chr.10: 14819936-14819936 on Build GRCh38
rs565190838 Chr.10: 14819898-14819898 on Build GRCh38
rs750397139 Chr.10: 14820002-14820002 on Build GRCh38
rs774620932 Chr.10: 14820064-14820064 on Build GRCh38
rs374288522 Chr.10: 14820041-14820041 on Build GRCh38
rs778707113 Chr.10: 14820094-14820094 on Build GRCh38

View Full Product Details