Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 73201195-73201436 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COG1
Entrez Gene ID
9382
Gene Name
component of oligomeric golgi complex 1
Gene Aliases
CDG2G, LDLB
UniGene
Hs.103555
Gene Chromosome Location
Chr.17: 73193034-73208507 on Build GRCh38
Transcripts
NM_018714.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6632181 c.1484G>A p.R495Q Chr.17: 73201311-73201311 on Build GRCh38
3820428 c.1525G>C p.A509P Chr.17: 73201352-73201352 on Build GRCh38
160106 c.1516A>G p.K506E Chr.17: 73201343-73201343 on Build GRCh38
4968820 c.1445A>G p.D482G Chr.17: 73201272-73201272 on Build GRCh38
6632186 c.1455C>T p.S485S Chr.17: 73201282-73201282 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1551036 Chr.17: 73201300-73201300 on Build GRCh38
rs927686295 Chr.17: 73201345-73201345 on Build GRCh38
rs565847659 Chr.17: 73201301-73201301 on Build GRCh38
rs138661086 Chr.17: 73201357-73201357 on Build GRCh38
rs773233123 Chr.17: 73201259-73201259 on Build GRCh38
rs769652614 Chr.17: 73201326-73201326 on Build GRCh38
rs759490050 Chr.17: 73201261-73201261 on Build GRCh38
rs934989828 Chr.17: 73201368-73201368 on Build GRCh38
rs191238196 Chr.17: 73201273-73201273 on Build GRCh38
rs760755764 Chr.17: 73201365-73201365 on Build GRCh38
rs780897656 Chr.17: 73201282-73201282 on Build GRCh38
rs149030357 Chr.17: 73201311-73201311 on Build GRCh38
rs535998327 Chr.17: 73201314-73201314 on Build GRCh38
rs199888646 Chr.17: 73201288-73201288 on Build GRCh38
rs372549949 Chr.17: 73201242-73201242 on Build GRCh38
rs746583075 Chr.17: 73201310-73201310 on Build GRCh38
rs141185616 Chr.17: 73201364-73201364 on Build GRCh38
rs780996847 Chr.17: 73201324-73201324 on Build GRCh38
rs762030507 Chr.17: 73201401-73201401 on Build GRCh38
rs34358576 Chr.17: 73201372-73201373 on Build GRCh38
rs201842059 Chr.17: 73201330-73201330 on Build GRCh38
rs554671974 Chr.17: 73201328-73201328 on Build GRCh38
rs764355806 Chr.17: 73201268-73201268 on Build GRCh38
rs770925715 Chr.17: 73201331-73201331 on Build GRCh38
rs143051453 Chr.17: 73201325-73201325 on Build GRCh38
rs150055852 Chr.17: 73201283-73201283 on Build GRCh38
rs764231325 Chr.17: 73201383-73201383 on Build GRCh38
rs543395876 Chr.17: 73201340-73201340 on Build GRCh38
rs779677895 Chr.17: 73201307-73201307 on Build GRCh38
rs768462230 Chr.17: 73201222-73201222 on Build GRCh38
rs962861585 Chr.17: 73201281-73201281 on Build GRCh38
rs1017397264 Chr.17: 73201252-73201252 on Build GRCh38
rs113486038 Chr.17: 73201243-73201243 on Build GRCh38
rs754090807 Chr.17: 73201279-73201279 on Build GRCh38
rs916184198 Chr.17: 73201287-73201287 on Build GRCh38
rs765613278 Chr.17: 73201406-73201406 on Build GRCh38
rs904722460 Chr.17: 73201385-73201385 on Build GRCh38
rs752817529 Chr.17: 73201265-73201265 on Build GRCh38
rs35401348 Chr.17: 73201221-73201222 on Build GRCh38
rs767566065 Chr.17: 73201263-73201263 on Build GRCh38
rs560931577 Chr.17: 73201276-73201276 on Build GRCh38
rs756327477 Chr.17: 73201267-73201267 on Build GRCh38
rs776561960 Chr.17: 73201232-73201232 on Build GRCh38

View Full Product Details