Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 39521237-39521496 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATF4
Entrez Gene ID
468
Gene Name
activating transcription factor 4
Gene Aliases
CREB-2, CREB2, TAXREB67, TXREB
UniGene
Hs.496487
Gene Chromosome Location
Chr.22: 39519709-39522686 on Build GRCh38
Transcripts
NM_001675.4 NM_182810.2 XM_017028807.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
726272 c.31G>C p.V11L Chr.22: 39521476-39521476 on Build GRCh38
1632598 c.1A>G p.M1V Chr.22: 39521446-39521446 on Build GRCh38
6390898 c.35T>A p.L12* Chr.22: 39521480-39521480 on Build GRCh38
4582419 c.40G>T p.G14W Chr.22: 39521485-39521485 on Build GRCh38
7126152 c.21G>C p.L7L Chr.22: 39521466-39521466 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs151149967 Chr.22: 39521345-39521345 on Build GRCh38
rs756538551 Chr.22: 39521403-39521403 on Build GRCh38
rs111935226 Chr.22: 39521353-39521353 on Build GRCh38
rs1001545052 Chr.22: 39521413-39521413 on Build GRCh38
rs751836565 Chr.22: 39521395-39521395 on Build GRCh38
rs779393729 Chr.22: 39521423-39521423 on Build GRCh38
rs768004395 Chr.22: 39521401-39521401 on Build GRCh38
rs367738767 Chr.22: 39521328-39521328 on Build GRCh38
rs371466949 Chr.22: 39521402-39521402 on Build GRCh38
rs902767391 Chr.22: 39521296-39521296 on Build GRCh38
rs1032908283 Chr.22: 39521312-39521312 on Build GRCh38
rs759839365 Chr.22: 39521448-39521448 on Build GRCh38
rs895305048 Chr.22: 39521420-39521420 on Build GRCh38
rs139077616 Chr.22: 39521369-39521369 on Build GRCh38
rs530230780 Chr.22: 39521432-39521432 on Build GRCh38
rs181328727 Chr.22: 39521349-39521349 on Build GRCh38
rs1054005603 Chr.22: 39521417-39521417 on Build GRCh38
rs1009820191 Chr.22: 39521287-39521287 on Build GRCh38
rs201201452 Chr.22: 39521446-39521446 on Build GRCh38
rs374660871 Chr.22: 39521421-39521421 on Build GRCh38
rs942515805 Chr.22: 39521367-39521367 on Build GRCh38
rs746700301 Chr.22: 39521428-39521428 on Build GRCh38
rs779630800 Chr.22: 39521471-39521471 on Build GRCh38
rs73885241 Chr.22: 39521295-39521295 on Build GRCh38
rs764339006 Chr.22: 39521463-39521463 on Build GRCh38
rs762759423 Chr.22: 39521440-39521440 on Build GRCh38
rs74751854 Chr.22: 39521314-39521314 on Build GRCh38
rs1040741627 Chr.22: 39521280-39521280 on Build GRCh38
rs759741126 Chr.22: 39521305-39521305 on Build GRCh38
rs146354334 Chr.22: 39521449-39521449 on Build GRCh38
rs189837769 Chr.22: 39521259-39521259 on Build GRCh38
rs371528882 Chr.22: 39521336-39521336 on Build GRCh38
rs970788338 Chr.22: 39521424-39521424 on Build GRCh38
rs11556097 Chr.22: 39521431-39521431 on Build GRCh38
rs900855773 Chr.22: 39521284-39521284 on Build GRCh38
rs751006399 Chr.22: 39521472-39521472 on Build GRCh38
rs972720499 Chr.22: 39521329-39521329 on Build GRCh38
rs945751296 Chr.22: 39521392-39521392 on Build GRCh38
rs372377952 Chr.22: 39521306-39521306 on Build GRCh38
rs369962392 Chr.22: 39521437-39521437 on Build GRCh38
rs140650335 Chr.22: 39521278-39521278 on Build GRCh38
rs942463488 Chr.22: 39521262-39521262 on Build GRCh38
rs755200555 Chr.22: 39521400-39521400 on Build GRCh38
rs778398197 Chr.22: 39521419-39521419 on Build GRCh38
rs544809451 Chr.22: 39521362-39521362 on Build GRCh38
rs757658895 Chr.22: 39521468-39521468 on Build GRCh38
rs149908720 Chr.22: 39521452-39521452 on Build GRCh38
rs1001072408 Chr.22: 39521307-39521307 on Build GRCh38
rs563469938 Chr.22: 39521319-39521319 on Build GRCh38
rs768241118 Chr.22: 39521429-39521429 on Build GRCh38
rs577428565 Chr.22: 39521358-39521358 on Build GRCh38
rs769564982 Chr.22: 39521433-39521433 on Build GRCh38
rs144184225 Chr.22: 39521320-39521320 on Build GRCh38
rs754403045 Chr.22: 39521467-39521467 on Build GRCh38
rs376351770 Chr.22: 39521441-39521441 on Build GRCh38

View Full Product Details