Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 118961792-118962023 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM184A
Entrez Gene ID
79632
Gene Name
family with sequence similarity 184 member A
Gene Aliases
C6orf60
UniGene
Hs.443789
Gene Chromosome Location
Chr.6: 118959829-119149193 on Build GRCh38
Transcripts
NM_001100411.2 NM_001288576.1 NM_024581.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6451999 c.3237C>T p.N1079N Chr.6: 118961865-118961865 on Build GRCh38
1072683 c.3271G>A p.D1091N Chr.6: 118961831-118961831 on Build GRCh38
4682783 c.3218T>C p.V1073A Chr.6: 118961884-118961884 on Build GRCh38
483340 c.3147G>T p.K1049N Chr.6: 118961955-118961955 on Build GRCh38
7128010 c.3150G>T p.K1050N Chr.6: 118961952-118961952 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769079636 Chr.6: 118961961-118961961 on Build GRCh38
rs868327690 Chr.6: 118961966-118961966 on Build GRCh38
rs767415400 Chr.6: 118961918-118961918 on Build GRCh38
rs776327295 Chr.6: 118961852-118961852 on Build GRCh38
rs781471924 Chr.6: 118961969-118961969 on Build GRCh38
rs1015880310 Chr.6: 118961857-118961857 on Build GRCh38
rs772584052 Chr.6: 118961823-118961823 on Build GRCh38
rs1001766863 Chr.6: 118961873-118961873 on Build GRCh38
rs954165482 Chr.6: 118961892-118961892 on Build GRCh38
rs772705480 Chr.6: 118961992-118961992 on Build GRCh38
rs755104527 Chr.6: 118961950-118961952 on Build GRCh38
rs868063202 Chr.6: 118961819-118961819 on Build GRCh38
rs368976311 Chr.6: 118961864-118961864 on Build GRCh38
rs754115785 Chr.6: 118961921-118961921 on Build GRCh38
rs766375460 Chr.6: 118961904-118961904 on Build GRCh38
rs779157141 Chr.6: 118961940-118961940 on Build GRCh38
rs759460715 Chr.6: 118961908-118961908 on Build GRCh38
rs761452555 Chr.6: 118961860-118961860 on Build GRCh38
rs762827450 Chr.6: 118961893-118961893 on Build GRCh38
rs202036689 Chr.6: 118961976-118961976 on Build GRCh38
rs769394356 Chr.6: 118961990-118961990 on Build GRCh38
rs747818356 Chr.6: 118961980-118961980 on Build GRCh38
rs764735318 Chr.6: 118961863-118961863 on Build GRCh38
rs575022229 Chr.6: 118961831-118961831 on Build GRCh38
rs775748737 Chr.6: 118961827-118961827 on Build GRCh38
rs372394552 Chr.6: 118961928-118961928 on Build GRCh38
rs768989755 Chr.6: 118961832-118961832 on Build GRCh38
rs376040804 Chr.6: 118961952-118961952 on Build GRCh38
rs750478410 Chr.6: 118961942-118961942 on Build GRCh38
rs1048024751 Chr.6: 118961865-118961865 on Build GRCh38
rs751425737 Chr.6: 118961907-118961907 on Build GRCh38
rs770240059 Chr.6: 118961878-118961878 on Build GRCh38
rs1040807765 Chr.6: 118961817-118961817 on Build GRCh38
rs780660408 Chr.6: 118961958-118961958 on Build GRCh38
rs888102632 Chr.6: 118961870-118961870 on Build GRCh38

View Full Product Details