Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 28683544-28683809 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMEB1
Entrez Gene ID
10691
Gene Name
glucocorticoid modulatory element binding protein 1
Gene Aliases
P96PIF, PIF96
UniGene
Hs.632373
Gene Chromosome Location
Chr.1: 28668728-28715603 on Build GRCh38
Transcripts
NM_001319674.1 NM_006582.3 NM_024482.2 XM_011540518.2 XM_011540519.2 XM_011540521.2 XM_011540522.2 XM_017000087.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5502191 c.31G>A p.G11R Chr.1: 28683643-28683643 on Build GRCh38
6306688 c.40G>T p.V14F Chr.1: 28683652-28683652 on Build GRCh38
6671990 c.16G>A p.V6M Chr.1: 28683628-28683628 on Build GRCh38
5502190 c.31G>A p.G11R Chr.1: 28683643-28683643 on Build GRCh38
6126177 c.127G>T p.G43W Chr.1: 28683739-28683739 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762553222 Chr.1: 28683777-28683777 on Build GRCh38
rs749877038 Chr.1: 28683578-28683578 on Build GRCh38
rs201655477 Chr.1: 28683783-28683783 on Build GRCh38
rs753173640 Chr.1: 28683697-28683697 on Build GRCh38
rs183408560 Chr.1: 28683575-28683575 on Build GRCh38
rs779007378 Chr.1: 28683723-28683723 on Build GRCh38
rs975785914 Chr.1: 28683754-28683754 on Build GRCh38
rs762489763 Chr.1: 28683651-28683651 on Build GRCh38
rs749449960 Chr.1: 28683598-28683598 on Build GRCh38
rs770085420 Chr.1: 28683593-28683593 on Build GRCh38
rs1006364102 Chr.1: 28683681-28683681 on Build GRCh38
rs375751705 Chr.1: 28683733-28683733 on Build GRCh38
rs747529948 Chr.1: 28683587-28683587 on Build GRCh38
rs768365056 Chr.1: 28683618-28683618 on Build GRCh38
rs952263656 Chr.1: 28683676-28683676 on Build GRCh38
rs186987064 Chr.1: 28683592-28683592 on Build GRCh38
rs369332884 Chr.1: 28683620-28683620 on Build GRCh38
rs771107458 Chr.1: 28683605-28683605 on Build GRCh38
rs201454881 Chr.1: 28683612-28683612 on Build GRCh38
rs986465158 Chr.1: 28683603-28683603 on Build GRCh38
rs769546907 Chr.1: 28683767-28683767 on Build GRCh38
rs372749317 Chr.1: 28683699-28683699 on Build GRCh38
rs778184285 Chr.1: 28683705-28683705 on Build GRCh38
rs770934513 Chr.1: 28683720-28683720 on Build GRCh38
rs200053822 Chr.1: 28683655-28683655 on Build GRCh38
rs200622672 Chr.1: 28683642-28683642 on Build GRCh38
rs755448063 Chr.1: 28683688-28683688 on Build GRCh38
rs775335497 Chr.1: 28683608-28683608 on Build GRCh38
rs765623162 Chr.1: 28683647-28683647 on Build GRCh38
rs781581800 Chr.1: 28683591-28683591 on Build GRCh38
rs768579198 Chr.1: 28683737-28683737 on Build GRCh38
rs896700391 Chr.1: 28683628-28683628 on Build GRCh38
rs772874532 Chr.1: 28683774-28683774 on Build GRCh38
rs561289258 Chr.1: 28683590-28683590 on Build GRCh38
rs773557591 Chr.1: 28683594-28683594 on Build GRCh38
rs140686141 Chr.1: 28683738-28683738 on Build GRCh38
rs761663207 Chr.1: 28683759-28683759 on Build GRCh38
rs756860623 Chr.1: 28683572-28683572 on Build GRCh38
rs749495011 Chr.1: 28683709-28683709 on Build GRCh38
rs920129499 Chr.1: 28683661-28683661 on Build GRCh38
rs773615760 Chr.1: 28683778-28683778 on Build GRCh38
rs11557120 Chr.1: 28683653-28683653 on Build GRCh38
rs780492378 Chr.1: 28683579-28683579 on Build GRCh38
rs922939770 Chr.1: 28683776-28683776 on Build GRCh38
rs150937827 Chr.1: 28683635-28683635 on Build GRCh38
rs781425414 Chr.1: 28683694-28683694 on Build GRCh38

View Full Product Details