Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 101655654-101655927 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMCX2
Entrez Gene ID
9823
Gene Name
armadillo repeat containing, X-linked 2
Gene Aliases
ALEX2, GASP9
UniGene
Hs.48924
Gene Chromosome Location
Chr.X: 101655281-101659891 on Build GRCh38
Transcripts
NM_001282231.1 NM_014782.6 NM_177949.3 XM_005278109.1 XM_005278110.1 XM_005278111.1 XM_005278113.1 XM_005278114.1 XM_005278115.1 XM_005278116.1 XM_005278117.1 XM_011531071.1 XM_011531072.1 XM_017029987.1 XM_017029988.1 XM_017029989.1 XM_017029990.1 XM_017029991.1 XM_017029992.1 XM_017029993.1 XM_017029994.1 XM_017029995.1 XM_017029996.1 XM_017029997.1

SNP Detail

SNP ID Chromosome Location
rs781957214 Chr.X: 101655792-101655792 on Build GRCh38
rs782159485 Chr.X: 101655739-101655739 on Build GRCh38
rs781919454 Chr.X: 101655686-101655686 on Build GRCh38
rs782671204 Chr.X: 101655896-101655896 on Build GRCh38
rs1030550437 Chr.X: 101655866-101655866 on Build GRCh38
rs373539462 Chr.X: 101655874-101655874 on Build GRCh38
rs782075172 Chr.X: 101655801-101655801 on Build GRCh38
rs782182924 Chr.X: 101655885-101655885 on Build GRCh38
rs150821075 Chr.X: 101655753-101655753 on Build GRCh38
rs377656852 Chr.X: 101655811-101655811 on Build GRCh38
rs781832160 Chr.X: 101655870-101655870 on Build GRCh38
rs782004650 Chr.X: 101655757-101655757 on Build GRCh38
rs782113386 Chr.X: 101655774-101655774 on Build GRCh38
rs782523715 Chr.X: 101655848-101655848 on Build GRCh38
rs781906043 Chr.X: 101655839-101655839 on Build GRCh38
rs139275799 Chr.X: 101655786-101655786 on Build GRCh38
rs782551420 Chr.X: 101655888-101655888 on Build GRCh38
rs201149044 Chr.X: 101655850-101655850 on Build GRCh38

View Full Product Details