Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 231241039-231241292 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf131
Entrez Gene ID
128061
Gene Name
chromosome 1 open reading frame 131
Gene Aliases
-
UniGene
Hs.743464
Gene Chromosome Location
Chr.1: 231223763-231241187 on Build GRCh38
Transcripts
NM_001300830.1 NM_152379.3 NM_001316350.1 NM_014236.3 XM_005273313.4 XM_011544303.2 XM_011544304.2

Gene Symbol
GNPAT
Entrez Gene ID
8443
Gene Name
glyceronephosphate O-acyltransferase
Gene Aliases
DAP-AT, DAPAT, DHAPAT, RCDP2
UniGene
Hs.498028
Gene Chromosome Location
Chr.1: 231241173-231277973 on Build GRCh38
Transcripts
NM_001300830.1 NM_152379.3 NM_001316350.1 NM_014236.3 XM_005273313.4 XM_011544303.2 XM_011544304.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4856835 c.17C>T p.S6L Chr.1: 231241125-231241125 on Build GRCh38
905458 c.71G>T p.S24I Chr.1: 231241071-231241071 on Build GRCh38
6009874 c.99C>T p.L33L Chr.1: 231241043-231241043 on Build GRCh38
373522 c.74C>T p.S25F Chr.1: 231241068-231241068 on Build GRCh38
4567365 c.67_68CC>TT p.P23F Chr.1: 231241074-231241075 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs546114860 Chr.1: 231241243-231241243 on Build GRCh38
rs199596832 Chr.1: 231241160-231241160 on Build GRCh38
rs888448028 Chr.1: 231241143-231241147 on Build GRCh38
rs777874673 Chr.1: 231241105-231241105 on Build GRCh38
rs1045757649 Chr.1: 231241264-231241264 on Build GRCh38
rs568068971 Chr.1: 231241222-231241222 on Build GRCh38
rs1015275715 Chr.1: 231241208-231241208 on Build GRCh38
rs780362961 Chr.1: 231241156-231241156 on Build GRCh38
rs755337225 Chr.1: 231241191-231241191 on Build GRCh38
rs530297711 Chr.1: 231241184-231241184 on Build GRCh38
rs143615379 Chr.1: 231241099-231241099 on Build GRCh38
rs762365587 Chr.1: 231241132-231241132 on Build GRCh38
rs776170264 Chr.1: 231241116-231241116 on Build GRCh38
rs2274067 Chr.1: 231241060-231241060 on Build GRCh38
rs758753162 Chr.1: 231241189-231241189 on Build GRCh38
rs897206104 Chr.1: 231241128-231241128 on Build GRCh38
rs774088754 Chr.1: 231241168-231241168 on Build GRCh38
rs553247724 Chr.1: 231241247-231241247 on Build GRCh38
rs746256762 Chr.1: 231241112-231241112 on Build GRCh38
rs750505380 Chr.1: 231241095-231241099 on Build GRCh38
rs554711764 Chr.1: 231241152-231241152 on Build GRCh38
rs531305779 Chr.1: 231241125-231241125 on Build GRCh38
rs985639179 Chr.1: 231241081-231241081 on Build GRCh38
rs777212154 Chr.1: 231241163-231241163 on Build GRCh38
rs757563319 Chr.1: 231241107-231241107 on Build GRCh38
rs961115285 Chr.1: 231241224-231241224 on Build GRCh38
rs957683173 Chr.1: 231241229-231241229 on Build GRCh38
rs753666241 Chr.1: 231241180-231241180 on Build GRCh38
rs542067193 Chr.1: 231241261-231241261 on Build GRCh38
rs936085903 Chr.1: 231241092-231241092 on Build GRCh38
rs758570090 Chr.1: 231241128-231241129 on Build GRCh38
rs752586434 Chr.1: 231241093-231241093 on Build GRCh38
rs754663350 Chr.1: 231241088-231241088 on Build GRCh38
rs1046487088 Chr.1: 231241127-231241127 on Build GRCh38
rs957746455 Chr.1: 231241245-231241245 on Build GRCh38
rs189779182 Chr.1: 231241145-231241145 on Build GRCh38
rs750575005 Chr.1: 231241119-231241130 on Build GRCh38
rs949990554 Chr.1: 231241134-231241134 on Build GRCh38
rs754960864 Chr.1: 231241220-231241220 on Build GRCh38
rs748844683 Chr.1: 231241166-231241166 on Build GRCh38
rs552479895 Chr.1: 231241120-231241120 on Build GRCh38
rs111417006 Chr.1: 231241181-231241181 on Build GRCh38
rs781206586 Chr.1: 231241091-231241091 on Build GRCh38
rs201276335 Chr.1: 231241111-231241111 on Build GRCh38
rs1005400031 Chr.1: 231241200-231241200 on Build GRCh38
rs113752547 Chr.1: 231241234-231241234 on Build GRCh38
rs746037124 Chr.1: 231241157-231241157 on Build GRCh38
rs762716272 Chr.1: 231241071-231241071 on Build GRCh38
rs749354278 Chr.1: 231241106-231241106 on Build GRCh38
rs773821506 Chr.1: 231241143-231241143 on Build GRCh38
rs537119013 Chr.1: 231241226-231241226 on Build GRCh38
rs760393961 Chr.1: 231241175-231241175 on Build GRCh38
rs200823696 Chr.1: 231241150-231241150 on Build GRCh38
rs764797964 Chr.1: 231241062-231241062 on Build GRCh38
rs999223725 Chr.1: 231241187-231241191 on Build GRCh38
rs1045767393 Chr.1: 231241148-231241148 on Build GRCh38
rs766060174 Chr.1: 231241138-231241138 on Build GRCh38
rs764028315 Chr.1: 231241176-231241176 on Build GRCh38
rs767137008 Chr.1: 231241169-231241169 on Build GRCh38
rs775328419 Chr.1: 231241173-231241173 on Build GRCh38
rs747366254 Chr.1: 231241161-231241161 on Build GRCh38
rs150128431 Chr.1: 231241115-231241115 on Build GRCh38
rs767261631 Chr.1: 231241146-231241146 on Build GRCh38
rs747558165 Chr.1: 231241117-231241117 on Build GRCh38
rs753946557 Chr.1: 231241167-231241167 on Build GRCh38
rs954292961 Chr.1: 231241233-231241233 on Build GRCh38
rs779217033 Chr.1: 231241156-231241156 on Build GRCh38
rs950145264 Chr.1: 231241259-231241259 on Build GRCh38
rs544457818 Chr.1: 231241130-231241130 on Build GRCh38
rs562281255 Chr.1: 231241269-231241269 on Build GRCh38
rs751437541 Chr.1: 231241083-231241083 on Build GRCh38
rs927304241 Chr.1: 231241270-231241270 on Build GRCh38
rs369068693 Chr.1: 231241073-231241073 on Build GRCh38
rs556696136 Chr.1: 231241230-231241230 on Build GRCh38
rs573141457 Chr.1: 231241258-231241258 on Build GRCh38
rs868789232 Chr.1: 231241232-231241232 on Build GRCh38
rs749920950 Chr.1: 231241066-231241066 on Build GRCh38
rs751977760 Chr.1: 231241190-231241190 on Build GRCh38

View Full Product Details