Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 63281198-63281459 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARFGAP1
Entrez Gene ID
55738
Gene Name
ADP ribosylation factor GTPase activating protein 1
Gene Aliases
ARF1GAP, HRIHFB2281
UniGene
Hs.25584
Gene Chromosome Location
Chr.20: 63272785-63289793 on Build GRCh38
Transcripts
NM_001281482.1 NM_001281483.1 NM_001281484.1 NM_018209.3 NM_175609.2 XM_006723823.2 XM_011528901.1 XM_017027950.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6602871 c.684+1G>A p.? Chr.20: 63281348-63281348 on Build GRCh38
1632392 c.684+1G>T p.? Chr.20: 63281348-63281348 on Build GRCh38
6602872 c.684+1G>A p.? Chr.20: 63281348-63281348 on Build GRCh38
7356096 c.658C>T p.R220W Chr.20: 63281321-63281321 on Build GRCh38
1632391 c.684+1G>T p.? Chr.20: 63281348-63281348 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762837725 Chr.20: 63281348-63281348 on Build GRCh38
rs6011718 Chr.20: 63281240-63281240 on Build GRCh38
rs760854686 Chr.20: 63281279-63281279 on Build GRCh38
rs571333981 Chr.20: 63281361-63281361 on Build GRCh38
rs779473844 Chr.20: 63281308-63281308 on Build GRCh38
rs556903155 Chr.20: 63281301-63281301 on Build GRCh38
rs767552979 Chr.20: 63281266-63281266 on Build GRCh38
rs770879609 Chr.20: 63281353-63281353 on Build GRCh38
rs535565023 Chr.20: 63281264-63281264 on Build GRCh38
rs769674209 Chr.20: 63281252-63281252 on Build GRCh38
rs752790290 Chr.20: 63281271-63281271 on Build GRCh38
rs751948056 Chr.20: 63281272-63281273 on Build GRCh38
rs769726600 Chr.20: 63281331-63281331 on Build GRCh38
rs750561446 Chr.20: 63281228-63281228 on Build GRCh38
rs1049867484 Chr.20: 63281392-63281392 on Build GRCh38
rs779188595 Chr.20: 63281291-63281291 on Build GRCh38
rs991248390 Chr.20: 63281310-63281310 on Build GRCh38
rs2236195 Chr.20: 63281363-63281363 on Build GRCh38
rs760839101 Chr.20: 63281354-63281354 on Build GRCh38
rs6122111 Chr.20: 63281347-63281347 on Build GRCh38
rs143690557 Chr.20: 63281235-63281235 on Build GRCh38
rs1008303938 Chr.20: 63281404-63281404 on Build GRCh38
rs374925495 Chr.20: 63281305-63281305 on Build GRCh38
rs1027964040 Chr.20: 63281236-63281236 on Build GRCh38
rs920042095 Chr.20: 63281277-63281277 on Build GRCh38
rs6011717 Chr.20: 63281225-63281225 on Build GRCh38
rs762004853 Chr.20: 63281359-63281359 on Build GRCh38
rs386816123 Chr.20: 63281361-63281363 on Build GRCh38
rs367622454 Chr.20: 63281362-63281362 on Build GRCh38
rs532994778 Chr.20: 63281406-63281406 on Build GRCh38
rs534137278 Chr.20: 63281391-63281391 on Build GRCh38
rs963142693 Chr.20: 63281417-63281417 on Build GRCh38
rs770590862 Chr.20: 63281396-63281396 on Build GRCh38
rs540004462 Chr.20: 63281375-63281375 on Build GRCh38
rs762121869 Chr.20: 63281340-63281342 on Build GRCh38
rs575263794 Chr.20: 63281319-63281319 on Build GRCh38
rs200990873 Chr.20: 63281389-63281389 on Build GRCh38
rs775389561 Chr.20: 63281273-63281273 on Build GRCh38
rs766933574 Chr.20: 63281364-63281364 on Build GRCh38
rs201153591 Chr.20: 63281384-63281384 on Build GRCh38
rs764321006 Chr.20: 63281280-63281280 on Build GRCh38
rs766315493 Chr.20: 63281374-63281374 on Build GRCh38
rs768235253 Chr.20: 63281320-63281320 on Build GRCh38
rs931397489 Chr.20: 63281306-63281306 on Build GRCh38
rs757619038 Chr.20: 63281288-63281288 on Build GRCh38
rs151072181 Chr.20: 63281411-63281411 on Build GRCh38
rs773127858 Chr.20: 63281341-63281341 on Build GRCh38
rs539331372 Chr.20: 63281321-63281321 on Build GRCh38
rs754018252 Chr.20: 63281285-63281285 on Build GRCh38
rs747737702 Chr.20: 63281322-63281322 on Build GRCh38
rs761726194 Chr.20: 63281242-63281242 on Build GRCh38
rs747868230 Chr.20: 63281386-63281386 on Build GRCh38
rs372002751 Chr.20: 63281393-63281393 on Build GRCh38
rs764091635 Chr.20: 63281358-63281358 on Build GRCh38
rs773289542 Chr.20: 63281253-63281253 on Build GRCh38
rs568416419 Chr.20: 63281239-63281239 on Build GRCh38
rs949677367 Chr.20: 63281332-63281332 on Build GRCh38

View Full Product Details