Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 72007074-72007307 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHST3
Entrez Gene ID
9469
Gene Name
carbohydrate sulfotransferase 3
Gene Aliases
C6ST, C6ST1, HSD
UniGene
Hs.158304
Gene Chromosome Location
Chr.10: 71964362-72013564 on Build GRCh38
Transcripts
NM_004273.4 XM_006718075.3 XM_011540369.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2161353 c.161A>G p.Q54R Chr.10: 72007192-72007192 on Build GRCh38
4836906 c.229C>T p.L77F Chr.10: 72007260-72007260 on Build GRCh38
7164340 c.156G>C p.L52L Chr.10: 72007187-72007187 on Build GRCh38
6587075 c.240G>A p.L80L Chr.10: 72007271-72007271 on Build GRCh38
920177 c.250G>A p.D84N Chr.10: 72007281-72007281 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766408307 Chr.10: 72007286-72007286 on Build GRCh38
rs267602570 Chr.10: 72007197-72007197 on Build GRCh38
rs1036460319 Chr.10: 72007175-72007175 on Build GRCh38
rs906323645 Chr.10: 72007117-72007117 on Build GRCh38
rs760211272 Chr.10: 72007230-72007230 on Build GRCh38
rs771240767 Chr.10: 72007223-72007223 on Build GRCh38
rs200494658 Chr.10: 72007208-72007208 on Build GRCh38
rs549789362 Chr.10: 72007272-72007272 on Build GRCh38
rs763906569 Chr.10: 72007126-72007126 on Build GRCh38
rs1046212726 Chr.10: 72007106-72007106 on Build GRCh38
rs749513659 Chr.10: 72007198-72007198 on Build GRCh38
rs760290305 Chr.10: 72007238-72007238 on Build GRCh38
rs758385149 Chr.10: 72007148-72007148 on Build GRCh38
rs995842020 Chr.10: 72007288-72007288 on Build GRCh38
rs142556853 Chr.10: 72007233-72007233 on Build GRCh38
rs1047318128 Chr.10: 72007101-72007101 on Build GRCh38
rs761589335 Chr.10: 72007255-72007255 on Build GRCh38
rs140362604 Chr.10: 72007206-72007206 on Build GRCh38
rs753475518 Chr.10: 72007253-72007253 on Build GRCh38
rs774717127 Chr.10: 72007225-72007225 on Build GRCh38
rs750283331 Chr.10: 72007268-72007268 on Build GRCh38
rs750348269 Chr.10: 72007146-72007146 on Build GRCh38
rs778856456 Chr.10: 72007145-72007145 on Build GRCh38
rs763709927 Chr.10: 72007245-72007245 on Build GRCh38
rs557708912 Chr.10: 72007187-72007187 on Build GRCh38
rs758673483 Chr.10: 72007215-72007215 on Build GRCh38
rs1008975704 Chr.10: 72007140-72007140 on Build GRCh38
rs748302738 Chr.10: 72007149-72007149 on Build GRCh38
rs964738146 Chr.10: 72007227-72007227 on Build GRCh38
rs759935618 Chr.10: 72007226-72007226 on Build GRCh38
rs773332801 Chr.10: 72007178-72007182 on Build GRCh38
rs753672743 Chr.10: 72007133-72007133 on Build GRCh38
rs765077860 Chr.10: 72007256-72007256 on Build GRCh38
rs375092999 Chr.10: 72007125-72007125 on Build GRCh38
rs766157121 Chr.10: 72007239-72007239 on Build GRCh38
rs778031221 Chr.10: 72007190-72007190 on Build GRCh38
rs375042671 Chr.10: 72007124-72007136 on Build GRCh38
rs368468468 Chr.10: 72007275-72007275 on Build GRCh38
rs757061413 Chr.10: 72007135-72007135 on Build GRCh38

View Full Product Details